Search
Buscar
View additional product information for Tantalum wire, 0.203mm (0.008in) dia, hard, 99.9% (metals basis) - FAQs (045133.G1, 045133.G6)
1 product FAQs found
"Hard" tantalum wire refers to the tensile strength, which is >1,000 Mpa.