Tantalum wire, 0.203mm (0.008in) dia, hard, 99.9% (metals basis)
Tantalum wire, 0.203mm (0.008in) dia, hard, 99.9% (metals basis)
Tantalum wire, 0.203mm (0.008in) dia, hard, 99.9% (metals basis)
Thermo Scientific Chemicals

Tantalum wire, 0.203mm (0.008in) dia, hard, 99.9% (metals basis)

CAS: 7440-25-7 | Ta | 180.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 m
10 m
Catalog number 045133.G6
also known as 045133-G6
Price (USD)
236.65
Special offer
Online exclusive
Ends: 28-Dec-2026
278.00
Save 41.35
Each
Quantity:
10 m
Request bulk or custom format
Price (USD)
236.65
Special offer
Online exclusive
Ends: 28-Dec-2026
278.00
Save 41.35
Each
Specifications
Boiling Point5429°C
CAS7440-25-7
ChEBICHEBI:33348
Chemical Name or MaterialTantalum wire
Density16.69
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Frequently asked questions (FAQs)

What does "hard" mean for the following tantalum wire product: Tantalum wire, 0.203 mm (0.008 in.) dia, hard, 99.9% (metals basis)?

"Hard" tantalum wire refers to the tensile strength, which is >1,000 Mpa.