Triton™ Series Multicollector Thermal Ionization Mass Spectrometer
Actual product may vary
Thermo Scientific™
Triton™ Series Multicollector Thermal Ionization Mass Spectrometer
Extract high-precision isotope ratio information from your samples with the Thermo Scientific™ Triton™ Series Multicollector Thermal Ionization Mass Spectrometer (TIMS).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeModel
0726203TIMSTriton XT
IQLAAEGAASFAFYMAMVTIMSTriton Plus
Catalog number 0726203
Price (MXN)
-
Type:
TIMS
Model:
Triton XT
Extract high-precision isotope ratio information from your samples with the Thermo Scientific™ Triton™ Series Multicollector Thermal Ionization Mass Spectrometer (TIMS).
The Thermo Scientific™ Triton™ XT TIMS integrates the established field-proven technologies from the Triton Series instruments with the latest developments in technology for isotope ratio analysis from Thermo Fisher Scientific™.
The result of over two decades of development
The Thermo Scientific™ Triton XT™ TIMS is the pinnacle of over two decades of continuous development, combining outstanding technologies such as:
  • 1013Ω Amplifier Technology: extending the lower limits of the ultimate detector
  • Retarding Potential Quadrupole Lenses (RPQ): for the highest abundance sensitivity allowing high precision of extreme isotope ratios
  • Multi-ion counting (MIC) packages: dedicated for specific nuclear and geochronology applications
  • Stable and linear SEM and then CDD ion counters: for the smallest sample amounts and lowest abundance isotopes
  • High-performance Faraday cups: to guarantee uniform response and long lifetime
  • High stability thermal ionization source: characterized by a very small kinetic energy spread of the ions and optimized for maximum ion transmission for both single and double filament techniques
  • Reliable moveable collector array: brings up to 9 Faraday cups into precise alignment with ion beams of different dispersions. With a mass range to cover static Li measurements, that is extendable to cover static Ca
  • Virtual amplifier: allows cancellation of gain factor uncertainties through amplifier rotation, so that low ppm external reproducibility can be achieved using efficient static measurements
Specifications
Includes1013 Ω amplifier technology with gain calibration and tau correction, central dual-channel detector (SEM/Faraday cup).
ModelTriton XT
TypeTIMS
Unit SizeEach