Search
View additional product information for Attune™ NxT and CytPix Small Particle Side-Scatter Filter - FAQs (100083194)
2 product FAQs found
When analyzing cells or large particles, you must use the standard SSC filter. If the Attune NxT Small Particle Side-Scatter Filter is used when analyzing cells or large particles, you may observe saturating signal.
The standard SSC filter has an OD 2 filter in addition to the 488/10 filter. The Attune NxT Small Particle Side-Scatter Filter does not include the OD 2 filter. This allows approximately 100-fold more light to get through the Attune NxT Small Particle Side-Scatter Filter.