Attune™ NxT and CytPix Small Particle Side-Scatter Filter
Invitrogen™
Attune™ NxT and CytPix Small Particle Side-Scatter Filter
The Attune NxT Small Particle Side-Scatter Filter is for use with the Attune NxT Acoustic Focusing Cytometer. It is designedRead more
Have Questions?
Catalog NumberQuantity
1000831941 filter
Catalog number 100083194
Price (USD)
449.00
Each
Quantity:
1 filter
Price (USD)
449.00
Each
The Attune NxT Small Particle Side-Scatter Filter is for use with the Attune NxT Acoustic Focusing Cytometer. It is designed to increase the dynamic range of the side-scatter detection without sacrificing resolution of the detector. The use of this 488/10 side scatter filter combined with ultra-filtering of the focusing fluid has been found to dramatically decrease background noise and enable discrimination of 100-nm particles.
Easy to use
This filter is easily swapped for the standard side-scatter filter. It cannot be installed the wrong way because the optical block is keyed to accept only one orientation of the filter blade. The filter also has a color-coded label to help identify the correct position.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Block FormatNon-Interchangeable
Product LineAttune
Quantity1 filter
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
For Use With (Equipment)Attune Acoustic Focusing Cytometer
Product TypeSmall Particle Side-Scatter Filter
Unit SizeEach
Frequently asked questions (FAQs)
Can I keep the Attune NxT Small Particle Side-Scatter Filter in place when analyzing cells or large particles, or must it be removed and replaced with the standard SSC filter?
When analyzing cells or large particles, you must use the standard SSC filter. If the Attune NxT Small Particle Side-Scatter Filter is used when analyzing cells or large particles, you may observe saturating signal.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
What is the difference between the Attune NxT Small Particle Side-Scatter Filter and the standard SSC filter?
The standard SSC filter has an OD 2 filter in addition to the 488/10 filter. The Attune NxT Small Particle Side-Scatter Filter does not include the OD 2 filter. This allows approximately 100-fold more light to get through the Attune NxT Small Particle Side-Scatter Filter.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.