Recombinant Protein G - Sepharose™ 4B, 2 mL - FAQs
Recombinant Protein G - Sepharose™ 4B, 2 mL - FAQs
View additional product information for Recombinant Protein G - Sepharose™ 4B - FAQs (101241, 101242, 101243)
1 product FAQs found
Can I perform a centrifugation separation with Recombinant Protein G - Sepharose 4B (Cat. Nos. 101241, 101242, 101243) instead of column separation?
Recombinant Protein G - Sepharose 4B (Cat. Nos. 101241, 101242, 101243) has not been validated for centrifugation separation.