Search
View additional product information for dNTP Set (100 mM) - FAQs (10297117, 10297018)
1 product FAQs found
Each deoxynucleotide (dATP, dCTP, dGTP, dTTP) is supplied at a concentration of 100 mM.