Search
View additional product information for RNase AWAY™ Decontamination Reagent - FAQs (10328011)
3 product FAQs found
There is no recommended time period for the RNase AWAY Decontamination Reagent (Cat. No. 10328011) to be in contact with the surface which has to be decontaminated. You do not need to leave the product act for a given time. Just apply it and rub with a wipe. After that you can rinse it with water.
Find additional tips, troubleshooting help, and resources within our RNA Sample Collection, Protection, and Isolation Support Center.
RNase AWAY Decontamination Reagent is a ready-to-use solution for eliminating RNase and DNA contamination from labware. Apply it evenly over the surface of glassware or plasticware to be treated and then rinse it away with distilled water.
RNase AWAY Decontamination Reagent is functionally tested for the elimination of RNase activity and DNA. No detectable RNase activity or DNA is observed.