Penicillin-Streptomycin-Glutamine (100X) - FAQs
Penicillin-Streptomycin-Glutamine (100X) - FAQs
View additional product information for Penicillin-Streptomycin-Glutamine (100X) - FAQs (10378016)
6 product FAQs found
What is the recommended concentration of Penicillin-Streptomycin-Glutamine to use?
The Penicillin-Streptomycin-Glutamine solution is at 100X. This can be used at a 1X concentration in media.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
There is a precipitate in my Penicillin-Streptomycin-Glutamine solution upon thaw, is this still good to use?
Yes, this is still good to use. The precipitate is due to the glutamine. The solution needs to be warmed to room temp and swirled unitl the glutamine goes back into solution.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
My Penicillin-Streptomycin-Glutamine solution has a color tint, is this normal?
Yes, this is normal and will not affect the potency or application of the product. This solution is typically colorless. However, it can have a pink to yellow color tint. The coloring is a carry-over from the manufacturing process of Streptomycin - the genus that Steptomycin is isolated from (Actinomycetes Streptomyces griseus) is responsible for a wide variety of pigments.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
For Penicillin-Streptomycin-Glutamine, what is the concentration of each component?
This contains: Penicillin G Sodium at 12,000 Units/mL, Streptomycin Sulfate at 10,000 mcg/mL, L-glutamine at 29.2 mg/mL (200 mM), and Sodium Citrate 0.14% NaCl at 10mM
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How can I decontaminate my cultures?
When an irreplaceable culture becomes contaminated, researchers may attempt to eliminate or control the contamination.
1. Determine if the contamination is bacteria, fungus, mycoplasma, or yeast. Read more here to view characteristics of each contaminant.
2. Isolate the contaminated culture from other cell lines.
3. Clean incubators and laminar flow hoods with a laboratory disinfectant, and check HEPA filters.
4. Antibiotics and antimycotics at high concentrations can be toxic to some cell lines. Therefore, perform a dose-response test to determine the level at which an antibiotic or antimycotic becomes toxic. This is particularly important when using an antimycotic such as Gibco Fungizone reagent or an antibiotic such as tylosin.
The following is a suggested procedure for determining toxicity levels and decontaminating cultures:
1. Dissociate, count, and dilute the cells in antibiotic-free media. Dilute the cells to the concentration used for regular cell passage.
2. Dispense the cell suspension into a multiwell culture plate or several small flasks. Add the antibiotic of choice to each well in a range of concentrations. For example, we suggest the following concentrations for Gibco Fungizone reagent: 0.25, 0.50, 1.0, 2.0, 4.0, and 8.0 µg/mL.
3. Observe the cells daily for signs of toxicity such as sloughing, appearance of vacuoles, decrease in confluency, and rounding.
4. When the toxic antibiotic level has been determined, culture the cells for two to three passages using the antibiotic at a concentration one- to two-fold lower than the toxic concentration.
5. Culture the cells for one passage in antibiotic-free media.
6. Repeat step 4.
7. Culture the cells in antibiotic-free medium for four to six passages to determine if the contamination has been eliminated.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What antibiotics do you offer to help control or eliminate cell culture contamination?
Please view the following page to browse the cell culture antibiotics we offer (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/antibiotics.html).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.