5-Aminoquinoline
5-Aminoquinoline
5-Aminoquinoline
Acros Organics

5-Aminoquinoline

CAS: 611-34-7 | C9H8N2 | 144.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 104600010
also known as 10460
HSN 29334900
Price (INR)
14,500.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
14,500.00
Each
Chemical Identifiers
CAS611-34-7
IUPAC Namequinolin-5-amine
Molecular FormulaC9H8N2
InChI KeyXMIAFAKRAAMSGX-UHFFFAOYSA-N
SMILESNC1=C2C=CC=NC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point106°C to 112°C
Appearance (Color)Yellow to brown
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only