Acide 2-benzoylbenzoïque, 98+ %, Thermo Scientific Chemicals
Acide 2-benzoylbenzoïque, 98+ %, Thermo Scientific Chemicals
Acide 2-benzoylbenzoïque, 98+ %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acide 2-benzoylbenzoïque, 98+ %, Thermo Scientific Chemicals

CAS: 85-52-9 | C14H10O3 | 226.23 g/mol
Have Questions?
Modifier l'affichagebuttonViewtableView
Quantité:
25 g
500 g
1 kg
Conditionnement:
Flacon en plastique
Référence 105710010
Prix (EUR)
214,00
Each
Quantité:
1 kg
Conditionnement:
Flacon en plastique
Grand volume ou format personnalisé
Prix (EUR)
214,00
Each
Identifiants chimiques
CAS85-52-9
Titration with NaOH>=98.0 %
Appearance (Color)White to light beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point126°C to 130°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only