Phytohemagglutinin, M form (PHA-M) - FAQs
View additional product information for Phytohemagglutinin, M form (PHA-M) - FAQs (10576015)
2 product FAQs found
What is the concentration of the PHA-M solution?
The PHA is a crude extract of red kidney beans. No protein concentration assay is performed. We recommend using the product at 1-2% by volume, or you may choose to optimize the dose by titration.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
For how long is Phytohemagglutinin, M form (PHA-M) stable one thawed. Can I freeze it again?
PHA-M is shipped on dry ice and recommended storage is at -5 degrees C to -20 degrees C.
Post-thawing, the product should be used within 10-15 days. We do not recommend freezing it back.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.