Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This polyclonal antibody raised against full-length human VDAC1 protein can cross react with VDAC2.
Immunogen sequence: MAVPPTYAD LGKSARDVFT KGYGFGLIKL DLKTKSENGL EFTSSGSANT ETTKVTGSLE TKYRWTEYGL TFTEKWNTDN TLGTEITVED QLARGLKLTF DSSFSPNTGK KNAKIKTGYK REHINLGCDM DFDIAGPSIR GALVLGYEGW LAGYQMNFET AKSRVTQSNF AVGYKTDEFQ LHTNVNDGTE FGGSIYQKVN KKLETAVNLA WTAGNSNTRF GIAAKYQIDP DACFSAKVNN SSLIGLGYTQ TLKPGIKLTL SALLDGKNVN AGGHKLGLGL EFQA (1-283 aa encoded by BC008482)
This gene encodes a voltage-dependent anion channel protein that is a major component of the outer mitochondrial membrane. The encoded protein facilitates the exchange of metabolites and ions across the outer mitochondrial membrane and may regulate mitochondrial functions. This protein also forms channels in the plasma membrane and may be involved in transmembrane electron transport. Alternate splicing results in multiple transcript variants. Multiple pseudogenes of this gene are found on chromosomes 1, 2 3, 6, 9, 12, X and Y.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B36-VDAC; epididymis secretory sperm binding protein; mVDAC1; mVDAC5; Non-selective voltage-gated ion channel VDAC1; Non-selective voltage-gated ion channel VDAC2; Outer mitochondrial membrane protein porin 1; Outer mitochondrial membrane protein porin 2; Plasmalemmal porin; Porin 31HL; Porin 31HM; rVDAC1; sperm binding protein 1a; unnamed protein product; VDAC-1; VDAC-2; VDAC-5; VDAC-6; VDAC2; Voltage-dependent anion-selective channel protein 1; voltage-dependent anion-selective channel protein 2; Voltage-dependent anion-selective channel protein 5; Voltage-dependent anion-selective channel protein 6
Gene Aliases: AL033343; mVDAC2; mVDAC6; POR; PORIN; VDAC; VDAC-1; VDAC1; VDAC2; Vdac5; Vdac6
UniProt ID: (Human) P21796, (Human) P45880, (Mouse) Q60932, (Rat) Q9Z2L0, (Mouse) Q60930, (Rat) P81155
Entrez Gene ID: (Human) 7416, (Human) 7417, (Mouse) 22333, (Rat) 83529, (Mouse) 22334, (Rat) 83531
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support