UltraPure™ DNase/RNase-Free Distilled Water, 10 x 500 mL - FAQs
View additional product information for UltraPure™ DNase/RNase-Free Distilled Water - FAQs (10977015, 10977023)
3 product FAQs found
Does Thermo Fisher Scientific offer a protease-free DNase?
We do not test for protease activity as part of our QC but there is PMSF (a protease inhibitor) in the storage buffer. Furthermore, in the preparation of DNase I, we use a soybean trypsin inhibitor column to remove proteases.
What is the USP conductivity specification for UltraPure DNase/RNase-Free Distilled Water?
We perform daily testing of the water according to USP specifications. The USP specification is that conductivity be “not more than 1 micromho/cm”.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Do you offer DNase- and RNAse-free distilled water?
Yes, we offer UltraPure DNase/RNase-Free Distilled Water (Cat. No. 10977015 or 10977023).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.