Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • CD49f (Integrin alpha 6) Antibodies

          Invitrogen

          CD49f (Integrin alpha 6) Monoclonal Antibody (eBioGoH3 (GoH3)), FITC, eBioscience™

          56 Published Figures
          63 References
          View all (43) CD49f (Integrin alpha 6) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD49f (Integrin alpha 6) Monoclonal Antibody (eBioGoH3 (GoH3)), FITC, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (56)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 57

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD49f (Integrin alpha 6) Antibody (11-0495-80) in Flow

          Staining of normal human peripheral blood cells with 0.125 µg of Rat IgG2a K Isotype Control FITC (Product # 11-4321-42) (blue histogram) or 0.125 µg of Anti-Human/Mouse CD49f (Integrin alpha 6) FITC (purple histogram). Cells in the lymphocyte gate were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Western Blot, Flow Cytometry (WB, Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Flow Cytometry (Flow)
          CD49f (Integrin alpha 6) Antibody in Immunohistochemistry (IHC)
          CD49f (Integrin alpha 6) Antibody in Immunocytochemistry (ICC/IF)
          CD49f (Integrin alpha 6) Antibody in Immunohistochemistry (IHC)
          CD49f (Integrin alpha 6) Antibody in Immunohistochemistry (IHC)
          CD49f (Integrin alpha 6) Antibody in Immunohistochemistry (IHC)
          CD49f (Integrin alpha 6) Antibody in Neutralization (Neu)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          11-0495-80

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          -
          View 1 publication 1 publication

          Immunohistochemistry (IHC)

          -
          View 4 publications 4 publications

          Immunocytochemistry (ICC/IF)

          -
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          0.25 µg/test
          View 55 publications 55 publications

          Neutralization (Neu)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), FITC, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          eBioGoH3 (GoH3)

          Conjugate

          FITC FITC FITC
          • Unconjugated
          • Alexa Fluor 700 
          • APC 
          • APC-eFluor 780
          • Biotin 
          • eFluor 450
          • PE 
          • PE-Cyanine7
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 780
          • Request custom conjugation

          Excitation/Emission Max

          498/517 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          25 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11149121

          Product Specific Information

          Description: The eBioGoH3 monoclonal antibody reacts with mouse and human CD49f, also known as integrin alpha 6, very late activation antigen 6 (VLA-6 alpha chain), and platelet gpIc. CD49f is a 120 kD transmembrane protein. CD49f associates with CD29, the integrin beta 1 chain, to form the VLA-6 complex; CD49f also associates with CD104, the integrin beta 4 chain, to form the alpha 6 beta 4 complex. CD49f is expressed primarily on T cells, monocytes, platelets, epithelial and endothelial cells. CD49f expression has also been found on germinal center B cells. The eBioGoH3 antibody is cross-reactive to integrin alpha 6 on human, mouse and bovine cells. This antibody has also been reported to have functional activity in blocking the binding of integrin alpha 6 to laminin.

          Applications Reported: This GoH3 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This eBioGoH3 (GoH3) antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          CDf (Integrin alpha-6, ITGA6) is the integrin alpha chain alpha 6 protein. Integrins are integral cell-surface proteins composed of an alpha chain and a beta chain. A given chain may combine with multiple partners resulting in different integrins. For example, alpha 6 may combine with beta 4 in the integrin referred to as TSP180, or with beta 1 in the integrin VLA-6. Integrins are known to participate in cell adhesion as well as cell-surface mediated signaling. Two transcript variants encoding different isoforms have been found for CDf. Functionally, integrins are cell surface receptors that interact with the extracellular matrix and mediate intracellular signals that affect cellular shape, mobility, and progression through the cell cycle in response to the extracellular matrix. Integrin receptors are composed of alpha and beta subunits, and form structural and functional linkages between the ECM (extra-cellular matrix) and intracellular cytoskeletal linker proteins. Signaling mediated by Intergrin/ECM interactions are also integrated with cellular responses to growth factor signaling to regulate cellular proliferation, cytoskeletal reorganization and other responses necessary for cellular survival. CDf is predominantly expressed by epithelia, and plays a critical structural role in the hemidesmosome. More than 18 alpha and 8 beta subunits with numerous splice variant isoforms have been identified in mammals. Certain integrins can also bind to soluble ligands or to counter-receptors on adjacent cells, such as the intracellular adhesion molecules (ICAMs), resulting in aggregation of cells.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: CD49 antigen-like family member F; DKFZp686J01244; extracellular matrix receptor subunit; FLJ18737; integrin alpha6B; integrin, alpha 6; unnamed protein product; very late activation protein 6

          View more View less

          Gene Aliases: 5033401O05Rik; CD49f; ITGA6A; ITGA6B; JEB6; VLA-6

          View more View less

          UniProt ID: (Human) P23229, (Mouse) Q61739

          View more View less

          Entrez Gene ID: (Human) 3655, (Mouse) 16403

          View more View less

          Function(s)
          protein binding insulin-like growth factor I binding signaling receptor activity neuregulin binding cadherin binding integrin binding laminin binding macromolecular complex binding
          Process(es)
          cell-substrate junction assembly cell adhesion cell-matrix adhesion integrin-mediated signaling pathway ectodermal cell differentiation positive regulation of neuron projection development positive regulation of cell migration cell-substrate adhesion nail development positive regulation of apoptotic process positive regulation of GTPase activity skin morphogenesis positive regulation of transcription from RNA polymerase II promoter leukocyte migration cell-cell adhesion negative regulation of extrinsic apoptotic signaling pathway positive regulation of cell-substrate adhesion positive regulation of cell-cell adhesion cell adhesion mediated by integrin odontogenesis of dentin-containing tooth filopodium assembly brown fat cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-5d6d8bc979-kv9nx:80/100.66.72.150:80.
          git-commit: e71199a02a6c1449b2ac9ded8316d5734e8aa2de
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.62.0-2026.11.45-1.0