Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • c-Kit (CD117) Antibodies

          Invitrogen

          CD117 (c-Kit) Monoclonal Antibody (2B8), FITC, eBioscience™

          90 Published Figures
          106 References
          View all (197) c-Kit (CD117) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD117 (c-Kit) Monoclonal Antibody (2B8), FITC, eBioscience™

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (90)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 91

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD117 (c-Kit) Antibody (11-1171-81) in Flow

          Staining of mouse bone marrow cells with Anti-Human/Mouse CD45R (B220) PE (Product # 12-0452-82) and 0.25 µg of Rat IgG2b K Isotype Control FITC (Product # 11-4031-82) (left) or 0.25 µg of Anti-Mouse CD117 (c-Kit) FITC (right). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Western Blot, Flow Cytometry (WB, Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Immunocytochemistry (ICC/IF)
          CD117 (c-Kit) Antibody in Immunocytochemistry (ICC/IF)
          CD117 (c-Kit) Antibody in Immunocytochemistry (ICC/IF)
          CD117 (c-Kit) Antibody in Immunocytochemistry (ICC/IF)
          CD117 (c-Kit) Antibody in Immunocytochemistry (ICC/IF)
          CD117 (c-Kit) Antibody in Immunocytochemistry (ICC/IF)
          CD117 (c-Kit) Antibody in Immunohistochemistry (IHC)
          CD117 (c-Kit) Antibody in Immunohistochemistry (IHC)
          CD117 (c-Kit) Antibody in Immunohistochemistry (IHC)
          CD117 (c-Kit) Antibody in Immunohistochemistry (IHC)
          CD117 (c-Kit) Antibody in Immunohistochemistry (IHC)
          CD117 (c-Kit) Antibody in Immunohistochemistry (IHC)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          11-1171-81

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          -
          View 1 publication 1 publication

          Immunohistochemistry (IHC)

          -
          View 7 publications 7 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 6 publications 6 publications

          Flow Cytometry (Flow)

          0.5 µg/test
          View 90 publications 90 publications

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse, Pig

          Published species

          Fish, Gerbil, Mouse

          Host/Isotype

          Rat / IgG2b, kappa

          Recommended Isotype Control

          Rat IgG2b kappa Isotype Control (eB149/10H5), FITC, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          2B8

          Conjugate

          FITC FITC FITC
          • Unconjugated
          • APC 
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 563
          • Brilliant UV 615
          • Brilliant UV 661
          • Brilliant UV 737
          • Brilliant Violet 421
          • Brilliant Violet 480
          • Brilliant Violet 605
          • Brilliant Violet 650
          • Brilliant Violet 786
          • eFluor 450
          • PE 
          • PE-Cyanine5
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Super Bright 702
          • Super Bright 780
          • Request custom conjugation

          Excitation/Emission Max

          498/517 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_465185

          Product Specific Information

          Description: The 2B8 monoclonal antibody reacts with mouse CD117, also known as c-Kit receptor, Steel factor receptor, and stem cell factor receptor. A member of the tyrosine kinase receptor family, this 145-kDa molecule is expressed by a majority of hematopoietic progenitor cells characterized in the mouse bone marrow as a small subset of cells positive for Sca-1 and Thy1 (Thy1 low) and negative for lineage markers. The interaction of the mouse c-Kit receptor and steel factor promotes the proliferation and differentiation of hematopoietic progenitor cells. CD117 is also expressed by mast cells and plays a role in signaling and activation of these cells.

          Applications Reported: The 2B8 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This 2B8 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          KIT (c-KIT) is a proto-oncogene and a type 3 transmembrane receptor for MGF (mast cell growth factor, also known as stem cell factor). KIT was first identified as the cellular homolog of the feline sarcoma viral oncogene v-kit. KIT together with its ligand regulates growth and activation of a variety of hemopoietic and non-hemopoietic cells. Mutations in KIT are associated with gastrointestinal stromal tumors, mast cell disease, acute myelogenous leukemia, and piebaldism. Recently, deregulation of the KIT receptor TK by the prevalent activation loop mutation D816V has served as a focal point in therapeutic strategies aimed at curbing neoplastic mast cell growth. c-Kit is expressed in hematopoietic stem cells, germ cells, mast cells and gastrointestinal tract cajal cells. Upon binding of its ligand stem cell factor (SCF), c-kit dimerizes, resulting in receptor activation and autophosphorylation of various tyrosine residues including tyrosine 703 located on the cytoplasmic domain of the receptor. This modification allows docking of Grb2 and activation of the Ras/ERK signaling pathway. SCF/c-kit can activate multiple downstream signaling pathways including PI3K, PLC-gamma and JAK/STAT. c-kit receptor activation is essential for hematopoiesis, stem cell maintenance and gametogenesis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: belly-spot; c-kit; c-kit proto-oncogene protein; ckit; dominant spotting; Dominant white spotting; kit oncogene; proto-oncogene c-Kit; proto-oncogene tyrosine-protein kinase Kit; SCFR; spotted sterile male; Steel Factor Receptor; tyrosine kinase; tyrosine-protein kinase Kit; unnamed protein product

          View more View less

          Gene Aliases: Bs; c-KIT; CD117; Fdc; Gsfsco1; Gsfsco5; Gsfsow3; MGF; SCO1; SCO5; SOW3; Ssm; Tr-kit; W

          View more View less

          UniProt ID: (Pig) Q2HWD6, (Mouse) P05532

          View more View less

          Entrez Gene ID: (Pig) 396810, (Mouse) 16590

          View more View less

          Function(s)
          transmembrane receptor protein tyrosine kinase activity ATP binding cytokine binding metal ion binding protease binding protein kinase activity protein tyrosine kinase activity stem cell factor receptor activity protein binding growth factor binding enzyme binding SH2 domain binding protein homodimerization activity transmembrane signal receptor
          Process(es)
          activation of MAPK activity ovarian follicle development immature B cell differentiation dendritic cell cytokine production inflammatory response spermatogenesis regulation of cell shape peptidyl-tyrosine phosphorylation cytokine-mediated signaling pathway lamellipodium assembly T cell differentiation erythrocyte differentiation melanocyte differentiation actin cytoskeleton reorganization mast cell cytokine production embryonic hemopoiesis megakaryocyte development Fc receptor signaling pathway Kit signaling pathway erythropoietin-mediated signaling pathway mast cell degranulation pigmentation digestive tract development stem cell differentiation detection of mechanical stimulus involved in sensory perception of sound cell chemotaxis mast cell differentiation melanocyte migration melanocyte adhesion hematopoietic progenitor cell differentiation myeloid progenitor cell differentiation lymphoid progenitor cell differentiation mast cell chemotaxis myeloid leukocyte differentiation positive regulation of cytokine production involved in immune response positive regulation of dendritic cell cytokine production protein phosphorylation glycosphingolipid metabolic process chemotaxis response to stress transmembrane receptor protein tyrosine kinase signaling pathway germ cell development spermatid development positive regulation of cell proliferation germ cell migration gonad development visual learning male gonad development response to radiation positive regulation of gene expression cell migration actin cytoskeleton organization hemopoiesis cell differentiation B cell differentiation positive regulation of cell migration positive regulation of pseudopodium assembly positive regulation of mast cell cytokine production somatic stem cell population maintenance ectopic germ cell programmed cell death intracellular signal transduction hematopoietic stem cell migration regulation of cell proliferation positive regulation of tyrosine phosphorylation of STAT protein negative regulation of programmed cell death positive regulation of MAP kinase activity positive regulation of MAPK cascade tongue development leukocyte activation positive regulation of Notch signaling pathway positive regulation of JAK-STAT cascade response to cadmium ion protein autophosphorylation developmental pigmentation positive regulation of long-term neuronal synaptic plasticity epithelial cell proliferation regulation of developmental process positive regulation of pyloric antrum smooth muscle contraction positive regulation of colon smooth muscle contraction positive regulation of small intestine smooth muscle contraction positive regulation of vascular smooth muscle cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-79565875f6-kzhm8:80/100.66.77.38:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline