Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • LILRB2 (ILT4) Antibodies

          Invitrogen

          CD85d (ILT4) Monoclonal Antibody (27D6), FITC, eBioscience™

          4 References
          View all (25) LILRB2 (ILT4) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD85d (ILT4) Monoclonal Antibody (27D6), FITC, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          CD85d (ILT4) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD85d (ILT4) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD85d (ILT4) Antibody (11-5148-42) in Flow

          Staining of normal human peripheral blood cells with Rat IgM kappa Isotype Control FITC (Product # 11-4341-82) (open histogram) or Anti-Human CD85d (ILT4) FITC (filled histogram). Cells in the monocyte gate were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD85d (ILT4) Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          11-5148-42

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          5 µL (0.5 µg)/test
          View 4 publications 4 publications
          Product Specifications

          Species Reactivity

          Human

          Published species

          Human

          Host/Isotype

          Rat / IgM

          Recommended Isotype Control

          Rat IgM Isotype Control (eBRM), FITC, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          27D6

          Conjugate

          FITC FITC FITC
          • Functional Grade
          • Request custom conjugation

          Excitation/Emission Max

          498/517 nm View spectra spectra

          Form

          Liquid

          Concentration

          5 µL/Test

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2, with BSA

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10853660

          Product Specific Information

          Description: The monoclonal antibody 27D6 recognizes the CD85d protein also known as ILT4, LIR-2. It is a member of the ILT (immunoglobulin-like transcript)/ LIR (leuokocyte Ig-like receptor) MIR (monocytes Ig-like receptor) family. CD85d is a 110 kDa single transmembrane receptor containing 3 Ig-like domains and a long cytoplasmic domain containing ITIMs (immunoreceptor tyrosine-based inhibitory motifs). Expression is confined to monocytes, macrophages and dendritic cells. The ligand for CD85d is non-classical class I HLA-G, -B and G1. Upon ligand binding, tyrosine phosphatase SHP-1 interacts with the ITIM. Interaction of HLA-G with ILT4/CD85d is thought to contribute to tolerance during fetal implantation. Additionally it is the levels of ILT3 and ILT4 on DCs that suggests a role in organ allograft rejection.

          The monoclonal antibody 27D6 has been shown to block binding of ligand (HLA-G tetramer).

          Applications Reported: This 27D6 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This 27D6 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.5 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

          Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          CD85d, also known as LILRB2 or ILT4, is a member of the immunoglobulin-like transcript (ILT), leukocyte Ig-like receptor (LIR), and monocyte Ig-like receptor (MIR) family. It is a 110 kDa single transmembrane receptor with three Ig-like domains and a long cytoplasmic domain containing immunoreceptor tyrosine-based inhibitory motifs (ITIMs). CD85d is expressed on monocytes, macrophages, and dendritic cells. The receptor binds to non-classical class I molecules such as HLA-G, -B, and G1. Upon ligand binding, the tyrosine phosphatase SHP-1 interacts with the ITIMs, transducing a negative signal that inhibits immune response stimulation. This interaction is thought to contribute to tolerance during fetal implantation and plays a role in controlling inflammatory responses and cytotoxicity, helping to focus the immune response and limit autoreactivity. CD85d is also involved in organ allograft rejection, with the levels of ILT3 and ILT4 on dendritic cells suggesting its role in this process. The LILRB2 gene, located in a gene cluster at chromosomal region 19q13.4, encodes multiple transcript variants with different isoforms, highlighting its complexity and importance in immune regulation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: CD85 antigen-like family member D; CD85d antigen; Ig-like transcript 4; immunoglobulin superfamily; leucocyte Ig-like receptor B2; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 2; LIR2CD85D; MIR10LILRA6; monocyte/macrophage Ig-related receptor; monocyte/macrophage immunoglobulin-like receptor 10; myeloid inhibitory receptor 10; unnamed protein product

          View more View less

          Gene Aliases: CD85D; ILT-4; ILT4; LIR-2; LIR2; MIR-10; MIR10

          View more View less

          UniProt ID: (Human) Q8N423

          View more View less

          Entrez Gene ID: (Human) 10288

          View more View less

          Function(s)
          beta-amyloid binding protein binding protein phosphatase 1 binding MHC class Ib protein complex binding MHC class Ib protein binding inhibitory MHC class I receptor activity MHC class I protein binding protein homodimerization activity macromolecular complex binding cell adhesion molecule binding
          Process(es)
          negative regulation of antigen processing and presentation positive regulation of tolerance induction positive regulation of T cell tolerance induction immune response-inhibiting cell surface receptor signaling pathway Fc receptor mediated inhibitory signaling pathway immune response cellular defense response signal transduction cell surface receptor signaling pathway cell-cell signaling learning or memory positive regulation of interleukin-6 production heterotypic cell-cell adhesion positive regulation of T cell proliferation negative regulation of T cell proliferation positive regulation of regulatory T cell differentiation negative regulation of protein metabolic process negative regulation of calcium ion transport cellular response to lipopolysaccharide interleukin-10-mediated signaling pathway regulation of long-term synaptic potentiation positive regulation of long term synaptic depression negative regulation of postsynaptic density organization negative regulation of T cell costimulation regulation of dendritic cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-q2vkv:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline