Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MPGLSCRFY QHKFPEVEDV VMVNVRSIAE MGAYVSLLEY NNIEGMILLS ELSRRRIRSI NKLIRIGRNE CVVVIRVDKE KGYIDLSKRR VSPEEAIKCE DKFTKSKTVY SILRHVAEVL EYTKDEQLES LFQRTAWVFD DKYKRPGYGA YDAFKHAVSD PSILDSLDLN EDEREVLINN INRRLTPQAV KIRADIEVAC YGYEGIDAVK EALRAGLNCS TENMPIKINL IAPPRYVMTT TTLERTEGLS VLSQAMAVIK EKIEEKRGVF NVQMEPKVVT DTDETELARQ MERLERENAE VDGDDDAEEM EAKAED (1-315 aa encoded by B C002513)
EIF2a (Eukaryotic translation initiation factor 2A) is a heterotrimer composed of three subunits (alpha, beta, and gamma). This translation initiation factor drives binding of initiator methionyl-tRNA to the 40S ribosome in an AUG-dependent manner to form the 43S pre-initiation complex. The polypeptide can be phosphorylated by related protein kinases activated in response to stress. Phosphorylated EIF2A inhibits EIF2B activity and prevents guanine nucleotide exchange.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eIF-2-alpha; EIF-2A; EIF-2alpha; eIF2 alpha; eIF2-alpha; Eukaryotic translation initiation factor 2 subunit 1; Eukaryotic translation initiation factor 2 subunit alpha; eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa; eukaryotic translation initiation factor 2A; translational initiation factor eIF-2, alpha subunit; unnamed protein product
Gene Aliases: 0910001O23Rik; 2410026C18Rik; 35kDa; EIF-2; EIF-2A; EIF-2alpha; EIF2; EIF2A; eIF2alpha; EIF2S1
UniProt ID: (Human) P05198, (Rat) P68101, (Mouse) Q6ZWX6
Entrez Gene ID: (Human) 1965, (Rat) 54318, (Mouse) 13665
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support