What is the water solubility of 3,3'-Diaminobenzidine tetrahydrochloride hydrate (Cat. No. 112090250)?
3,3'-Diaminobenzidine tetrahydrochloride hydrate is soluble in water at 50 mg/ml.
Search
View additional product information for 3,3'-Diaminobenzidine tetrahydrochloride hydrate, 97%, contains up to 10% water - FAQs (112090250)
1 product FAQs found