AmnioMAX™-II Complete Medium - FAQs
View additional product information for AmnioMAX™-II Complete Medium - FAQs (11269016)
1 product FAQs found
Can I supplement AmnioMax-II Complete Medium (Cat. No. 11269016) with Fungizone?
No. We do not recommend supplementing AmnioMAX-II Complete Medium (Cat. No. 1129016) with Fungizone, as it can be detrimental to amniocyte growth.  
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.