1,8-Dibromooctane, 98%
1,8-Dibromooctane, 98%
1,8-Dibromooctane, 98%
Thermo Scientific Chemicals

1,8-Dibromooctane, 98%

CAS: 4549-32-0 | C8H16Br2 | 272.01 g/mol
数量:
10 mL
50 mL
250 mL
パッケージ:
すべて表示
Glass bottle
製品番号(カタログ番号) 112880500
価格(JPY)
-
数量:
50 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4549-32-0
IUPAC Name1,8-dibromooctane
Molecular FormulaC8H16Br2
InChI KeyDKEGCUDAFWNSSO-UHFFFAOYSA-N
SMILESBrCCCCCCCCBr
さらに表示
GC>=97.5 %
Refractive index1.4981 to 1.5001 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear yellow
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only