Chemically Defined Lipid Concentrate - FAQs
View additional product information for Chemically Defined Lipid Concentrate - FAQs (11905031)
1 product FAQs found
Do you have stability/degradation information for Chemically Defined Lipid Concentrate (Cat. No. 11905031)? How has the product been filtered?
The shelf life of the product in the closed bottle is 12 months from the date of manufacture. In order to prevent “air encounters”, we recommend keeping the head space in the bottle as small as possible. You may consider an Argon overlay as well. Argon being heavier than air will settle on top of the chemical. We recommend dividing the contents of the bottle (100 mL) in usable aliquots ideally such that each aliquot is used up in one application.
The product is filtered with a 0.22 µm Durapore filter or Cartridge from EMD Millipore (Cat. No. MSP000812).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.