Product Image
Gibco™

Chemically Defined Lipid Concentrate

Insect cell culture tested.Chemically Defined Lipid Concentrate is a concentrated lipid emulsion designed to reduce or replace fetal bovine serumRead more
Have Questions?
Catalog NumberQuantity
11905031100 mL
Catalog number 11905031
Price (USD)
113.00
Each
Quantity:
100 mL
Price (USD)
113.00
Each
Insect cell culture tested.

Chemically Defined Lipid Concentrate is a concentrated lipid emulsion designed to reduce or replace fetal bovine serum in cell culture media for a wide variety of applications, including growth and maintenance of CHO, hybridoma, and insect cells in culture; monoclonal antibody production by hybridomas; and viral expression in insect cells. The components of this medium supplement are chemically-defined.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeMammalian Cells
Quantity100 mL
Shelf Life12 Months
Shipping ConditionRoom Temperature
FormLiquid
Product TypeLipid Concentrate
pH4 to 6
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.

Frequently asked questions (FAQs)

Do you have stability/degradation information for Chemically Defined Lipid Concentrate (Cat. No. 11905031)? How has the product been filtered?

The shelf life of the product in the closed bottle is 12 months from the date of manufacture. In order to prevent “air encounters”, we recommend keeping the head space in the bottle as small as possible. You may consider an Argon overlay as well. Argon being heavier than air will settle on top of the chemical. We recommend dividing the contents of the bottle (100 mL) in usable aliquots ideally such that each aliquot is used up in one application.

The product is filtered with a 0.22 µm Durapore filter or Cartridge from EMD Millipore (Cat. No. MSP000812).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.