Search
View additional product information for SuperScript™ Double-Stranded cDNA Synthesis Kit - FAQs (11917020, 11917010)
1 product FAQs found
To generate double-stranded cDNA, we recommend the Maxima H Minus Double-Stranded cDNA Synthesis Kit (Cat. No. K2561) or Second Strand cDNA Synthesis Kit (Cat. No. A48570).