Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CXCR4 (CD184) Antibodies

          Invitrogen

          CD184 (CXCR4) Monoclonal Antibody (2B11), PE, eBioscience™

          18 Published Figures
          63 References
          View all (85) CXCR4 (CD184) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD184 (CXCR4) Monoclonal Antibody (2B11), PE, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (18)
          CD184 (CXCR4) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD184 (CXCR4) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 19

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD184 (CXCR4) Antibody (12-9991-81) in Flow

          Staining of BALB/c thymocytes with Anti-Mouse CD4 FITC (Product # 11-0041-82) and 0.5 µg of Rat IgG2b kappa Isotype Control PE (Product # 12-4031-82) (left) or 0.5 µg of Anti-Mouse CD184 (CXCR4) PE (right). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD184 (CXCR4) Antibody in Flow Cytometry (Flow)
          CD184 (CXCR4) Antibody in Flow Cytometry (Flow)
          CD184 (CXCR4) Antibody in Flow Cytometry (Flow)
          CD184 (CXCR4) Antibody in Flow Cytometry (Flow)
          CD184 (CXCR4) Antibody in Flow Cytometry (Flow)
          CD184 (CXCR4) Antibody in Flow Cytometry (Flow)
          CD184 (CXCR4) Antibody in Flow Cytometry (Flow)
          CD184 (CXCR4) Antibody in Flow Cytometry (Flow)
          CD184 (CXCR4) Antibody in Flow Cytometry (Flow)
          CD184 (CXCR4) Antibody in Flow Cytometry (Flow)
          CD184 (CXCR4) Antibody in Flow Cytometry (Flow)
          CD184 (CXCR4) Antibody in Flow Cytometry (Flow)
          CD184 (CXCR4) Antibody in Flow Cytometry (Flow)
          CD184 (CXCR4) Antibody in Flow Cytometry (Flow)
          CD184 (CXCR4) Antibody in Flow Cytometry (Flow)
          CD184 (CXCR4) Antibody in Flow Cytometry (Flow)
          CD184 (CXCR4) Antibody in Flow Cytometry (Flow)
          CD184 (CXCR4) Antibody in Flow Cytometry (Flow)
          CD184 (CXCR4) Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          12-9991-81

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          -
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          1 µg/test
          View 61 publications 61 publications

          ELISA (ELISA)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Hamster, Human, Mouse

          Host/Isotype

          Rat / IgG2b, kappa

          Recommended Isotype Control

          Rat IgG2b kappa Isotype Control (eB149/10H5), PE, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          2B11

          Conjugate

          PE PE PE
          • Unconjugated
          • Alexa Fluor 488 
          • APC 
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 805
          • eFluor 450
          • Functional Grade
          • PE-Cyanine5.5
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-eFluor 710
          • Super Bright 600
          • Request custom conjugation

          Excitation/Emission Max

          565/576 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          50 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_891393

          Product Specific Information

          Description: The 2B11 monoclonal antibody reacts with mouse and human CXCR4, also known as CD184, Fusin, LESTR, or HUMSTR. Fusin is a member of the G-protein-coupled chemokine receptor family with seven membrane-spanning domains, and functions as a coreceptor for X4 HIV-1 entry into CD4+ cells. In addition it has been shown to play a role in lymphocyte migration. The ligand for CXCR4 is SDF1. CXCR4 is required for maintenance of granulocyte progenitors in the bone marrow and essential for B cell development. Expression of CXCR4 is different between mice and human. In the thymus CXCR4 is restricted to CD4+CD8+ cells while in the spleen predominant expression is found on B lymphocytes.

          The monoclonal antibody 2B11 recognizes the amino terminus of CXCR4 and has been reported to have functional activity; it does not block SDF1a but causes internalization of the receptor.

          Applications Reported: This 2B11 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This 2B11 antibody has been tested by flow cytometric analysis of mouse thymocytes. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.It is recommended that the incubation time with 2B11 antibody by at least 45 minutes for optimal staining.

          Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          CXCR4 (Fusin, LESTR, HUMSTR) is a member of the G-protein-coupled chemokine receptor family with seven membrane-spanning domains, and functions as a co-receptor for X4 HIV-1 entry into CD4+ cells. CXCR4 has been proved to be the co-receptor for HIV-2's binding to CD4 through envelope glycoprotein gp 120 and promotes Env-mediated fusion of the virus. In other cases, CXCR4 can even function as the only receptor for HIV-2's binding to the CD4 host cells. CXCR4 is the receptor for the C-X-C chemokine CXCL12/SDF-1 that transduces a signal by increasing intracellular calcium ion levels and enhancing MAPK1/MAPK3 activation. CXCR4 acts as a receptor for extracellular ubiquitin; leading to enhanced intracellular calcium ions and reduced cellular cAMP levels. Also, CXCR4 is involved in hematopoiesis and in cardiac ventricular septum formation, and plays an essential role in vascularization of the gastrointestinal tract, probably by regulating vascular branching and/or remodeling processes in endothelial cells. In the CNS, CXCR4 could mediate hippocampal-neuron survival and is involved in cerebellar development. CXCR4 binds bacterial lipopolysaccharide (LPS) and mediates LPS-induced inflammatory response, including TNF secretion by monocytes. Antibodies to CXCR4 block HIV-1 and HIV-2 fusion and infection of human target cells. The amino-terminal domain and the second extracellular loop of CXCR4 serve as HIV biding sites.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; CD antigen CD184; CD184; LESTR; PB-CKR; chemokine (C-X-C motif) receptor 4; chemokine (C-X-C) receptor 4; chemokine receptor 4; CXC; CXC motif chemokine; fusin; HUMSTR; leukocyte-derived seven transmembrane domain receptor; leukocyte-expressed seven-transmembrane-domain; pre-B-cell-derived chemokine receptor; SDF-1 receptor; stromal cell-derived factor 1 receptor

          View more View less

          Gene Aliases: b2b220Clo; CD184; Cmkar4; CXC-R4; CXCR-4; LESTR; PB-CKR; PBSF/SDF-1; Sdf1r

          View more View less

          UniProt ID: (Mouse) P70658

          View more View less

          Entrez Gene ID: (Mouse) 12767

          View more View less

          Function(s)
          actin binding G-protein coupled receptor activity chemokine receptor activity protein binding C-C chemokine receptor activity C-X-C chemokine receptor activity cytokine binding C-C chemokine binding ubiquitin protein ligase binding myosin light chain binding small molecule binding C-X-C motif chemokine 12 receptor activity ubiquitin binding
          Process(es)
          patterning of blood vessels ameboidal-type cell migration neuron migration kidney development positive regulation of protein phosphorylation epithelial cell development ventricular septum development chemotaxis immune response G-protein coupled receptor signaling pathway adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway positive regulation of cytosolic calcium ion concentration germ cell development nervous system development brain development motor neuron axon guidance germ cell migration organ morphogenesis cell migration calcium-mediated signaling neurogenesis telencephalon cell migration regulation of cell adhesion regulation of cell migration positive regulation of cell migration positive regulation of vascular wound healing hematopoietic stem cell migration aorta development CXCL12-activated CXCR4 signaling pathway T cell proliferation regulation of programmed cell death myelin maintenance endothelial cell differentiation generation of neurons positive regulation of oligodendrocyte differentiation positive regulation of neurogenesis regulation of viral process regulation of chemotaxis positive regulation of chemotaxis detection of temperature stimulus involved in sensory perception of pain detection of mechanical stimulus involved in sensory perception of pain regulation of calcium ion transport cardiac muscle contraction cell chemotaxis endothelial tube morphogenesis neural precursor cell proliferation positive regulation of ERK1 and ERK2 cascade cellular response to cytokine stimulus positive regulation of calcium ion import positive regulation of cold-induced thermogenesis response to tacrolimus positive regulation of dendrite extension positive regulation of vasculature development positive regulation of mesenchymal stem cell migration positive regulation of macrophage migration inhibitory factor signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-7d6nq:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline