Search
View additional product information for DynaMag™- Spin Magnet - FAQs (12320D)
12 product FAQs found
The DynaMag-2 Magnet is 3,500-3,700 minimum Gauss level and the DynaMag-Spin is 3,000-3,500 minimum Gauss level.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
1 Tesla is equivalent to 10,000 Gauss. .
Spray and/or wipe the DynaMag magnet unit with one of the following cleaning agents:
- 70% (v/v) isopropyl alcohol
- 1% (v/v) sodium hypochlorite solution (bleach)
- 0.1 N HCl solution
Do not submerge the magnet unit in aqueous solutions and avoid prolonged exposure to water or aqueous solutions. Clean with a damp cloth and mild detergent when exposed to harsh solvents. Do not autoclave the magnet unit as heating the magnet to above 70 degrees C can destabilize the magnetic material, resulting in loss of magnetic strength.
The DynaMag-2 Magnet can hold 16 standard 1.5 mL or 2 mL microcentrifuge tubes, while the DynaMag-Spin Magnet holds up to 6 standard 1.5 mL microcentrifuge tubes.
Superparamagnetic means that the Dynabeads magnetic beads exhibit magnetic properties when placed within a magnetic field, but have no residual magnetism when removed from the magnetic field.
This means that your targeted cells, proteins, or nucleic acids are only subjected to magnetic forces during the time the beads are on the magnet. The beads do not aggregate, but remain evenly dispersed in suspension.
Find additional tips, troubleshooting help, and resources within our Dynabeads Cell Isolation and Expansion Support Center.
Yes. The antibodies are covalently bound and should be very stable in your applications.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Depending on the antibody coated on the Dynabeads magnetic beads, the shelf life can vary from 24-36 months.
Some kits may have 18 months shelf life depending on other components supplied in the kit. The kits are guaranteed for 6 months from when you receive them.
Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.
1 Tesla is equivalent to 10,000 Gauss
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Spray and/or wipe the DynaMag magnet unit with one of the following cleaning agents:
- 70% (v/v) isopropyl alcohol
- 1% (v/v) sodium hypochlorite solution (bleach)
- 0.1 N HCl solution
Do not submerge the magnet unit in aqueous solutions and avoid prolonged exposure to water or aqueous solutions. Clean with a damp cloth and mild detergent when exposed to harsh solvents. Do not autoclave the magnet unit as heating the magnet to above 70 degrees C can destabilize the magnetic material, resulting in loss of magnetic strength.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
The DynaMag-2 Magnet (Cat. No. 12321D) can hold 16 standard 1.5 mL or 2 mL microcentrifuge tubes, while the DynaMag-Spin Magnet (Cat. No. 12320D) holds up to 6 standard 1.5 mL microcentrifuge tubes.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Spray and/or wipe the DynaMag magnet unit with one of the following cleaning agents:
- 70% (v/v) isopropyl alcohol
- 1% (v/v) sodium hypochlorite solution (bleach)
- 0.1 N HCl solution
Do not submerge the magnet unit in aqueous solutions and avoid prolonged exposure to water or aqueous solutions. Clean with a damp cloth and mild detergent when exposed to harsh solvents. Do not autoclave the magnet unit as heating the magnet to above 70 degrees C can destabilize the magnetic material, resulting in loss of magnetic strength.
We recommend that you select a magnet based on your sample volume:
- Invitrogen DynaMag-2 Magnet (Cat. No. 12321D) magnet working volume: 1 µl-1.5 mL sample volume
- Invitrogen DynaMag-5 Magnet (Cat. No. 12303D) magnet working volume: 1-5 mL sample volume
- Invitrogen DynaMag-15 Magnet (Cat. No. 12301D) magnet working volume: 4-15 mL sample volume
- Invitrogen DynaMag-50 Magnet (Cat. No. 12302D) magnet working volume: 10-50 mL sample volume
- Invitrogen DynaMag-96 Side Magnet (Cat. No. 123-31D): pulls beads to the side of each well. Compatible with PCR-strips and 96-well plates (200µL), non-skirted and half-skirted
- Invitrogen DynaMag-96 Bottom Magnet (Cat. No. 123-32D): pulls beads to the bottom of each well. Compatible with PCR-strips and 96-well plates (200µL) non-skirted and half-skirted