Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • UBAP1 Antibodies

          Proteintech

          UBAP1 Polyclonal Antibody

          17 References
          View all (9) UBAP1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite UBAP1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (24)
          UBAP1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          UBAP1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 24

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          UBAP1 Antibody (12385-1-AP) in ICC/IF

          Immunofluorescent analysis of Hela cells, using UBAP1 antibody 12385-1-AP at 1:25 dilution and Rhodamine-labeled goat anti-rabbit IGG (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          UBAP1 Antibody in Immunocytochemistry (ICC/IF)
          UBAP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBAP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBAP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBAP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBAP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBAP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBAP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBAP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBAP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBAP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UBAP1 Antibody in Western Blot (WB)
          UBAP1 Antibody in Western Blot (WB)
          UBAP1 Antibody in Western Blot (WB)
          UBAP1 Antibody in Western Blot (WB)
          UBAP1 Antibody in Western Blot (WB)
          UBAP1 Antibody in Western Blot (WB)
          UBAP1 Antibody in Western Blot (WB)
          UBAP1 Antibody in Western Blot (WB)
          UBAP1 Antibody in Western Blot (WB)
          UBAP1 Antibody in Western Blot (WB)
          UBAP1 Antibody in Western Blot (WB)
          UBAP1 Antibody in Western Blot (WB)
          UBAP1 Antibody in Immunoprecipitation (IP)
          UBAP1 Polyclonal Antibody

          Product Details

          12385-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2400
          View 15 publications 15 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          UBAP1 Fusion Protein Ag3055 (203-502 aa encoded by BC020950)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          30 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: GGSGSVLQD EEVLASLERA TLDFKPLHKP NGFITLPQLG NCEKMSLSSK VSLPPIPAVS NIKSLSFPKL DSDDSNQKTA KLASTFHSTS CLRNGTFQNS LKPSTQSSAS ELNGHHTLGL SALNLDSGTE MPALTSSQMP SLSVLSVCTE ESSPPNTGPT VTPPNFSVSQ VPNMPSCPQA YSELQMLSPS ERQCVETVVN MGYSYECVLR AMKKKGENIE QILDYLFAHG QLCEKGFDPL LVEEALEMHQ CSEEKMMEFL QLMSKFKEMG FELKDIKEVL LLHNNDQDNA LEDLMARAGA S (203-502 aa encoded by BC020950)

          Target Information

          Originally named because of its sequence similarity to the Saccharomyces cerevisiae NHP2 (non-histone protein 2), this protein appears to be a highly conserved nuclear protein that is a component of the [U4/U6.U5] tri-snRNP. It binds to the 5' stem-loop of U4 snRNA. Two transcript variants encoding the same protein have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Nasopharyngeal carcinoma-associated gene 20 protein; UAP; UBAP; UBAP-1; UBAP1; Ubiquitin-associated protein 1; unnamed protein product

          View more View less

          Gene Aliases: 2700092A01Rik; NAG20; SPG80; UAP; UBAP; UBAP-1; UBAP1

          View more View less

          UniProt ID: (Human) Q9NZ09, (Rat) Q5XIS7, (Mouse) Q8BH48

          View more View less

          Entrez Gene ID: (Human) 51271, (Rat) 362502, (Mouse) 67123

          View more View less

          Function(s)
          protein binding ubiquitin binding
          Process(es)
          protein transport ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway transport multivesicular body assembly protein targeting to vacuole involved in ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway membrane fission
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline