Are NP40 and DTT required for SUMO Protease activity? Will they interfere with downstream applications?
Yes, they are both required, although it might be possible to substitute the NP40 with another detergent via dialysis if you have concern about interference with mass spectrometry. DTT is an absolute requirement.