Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: VEFFRVVSE YRASLQQRMN PKQEEHAFES PPPPDVGGKD GLMDSTPALT PTEDLTPGSV EEAEEAEPDE EFKDAIEETH LVVLEDDEGA PCPAEDELAL QDNGFLSKNE VLRSKVSRLT ERLRKRYPTN NFGNCTGCSA TFSVLKKRRS CSNCGNSFCS RCCSFKVPKS SMGATAPEAQ RETVFVCASC NQTLSK (217-411 aa encoded by BC030621)
This gene encodes a protein with several transmembrane domains, a Rab11-binding domain and a lipid-binding FYVE finger domain. The encoded protein appears to promote neurite formation. A mutation in this gene has been reported to be associated with hereditary spastic paraplegia, however the pathogenicity of the mutation, which may simply represent a polymorphism, is unclear.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protrudin; RP11-459F3.2; Spastic paraplegia 33 protein; unnamed protein product; zinc finger FYVE domain containing 27; Zinc finger FYVE domain-containing protein 27
Gene Aliases: 2210011N02Rik; 9530077C24Rik; AI426636; AI593546; AI835681; PROTRUDIN; SPG33; ZFYVE27
UniProt ID: (Human) Q5T4F4, (Rat) Q6P7B7, (Mouse) Q3TXX3
Entrez Gene ID: (Human) 118813, (Rat) 309376, (Mouse) 319740
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support