Methyl sulfide, 99+%, extra pure
Methyl sulfide, 99+%, extra pure
Methyl sulfide, 99+%, extra pure
Thermo Scientific Chemicals

Methyl sulfide, 99+%, extra pure

CAS: 75-18-3 | C2H6S | 62.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 127760100
Price (USD)
41.00
Each
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
41.00
Each
Chemical Identifiers
CAS75-18-3
IUPAC Name(methylsulfanyl)methane
Molecular FormulaC2H6S
InChI KeyQMMFVYPAHWMCMS-UHFFFAOYSA-N
SMILESCSC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Water=<0.3 %
Refractive index1.4341 to 1.4361 (20°C, 589 nm)
GC=<0.2 % Methylmercaptan
Appearance (Color)Clear colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only