Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • New Products
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • FOXP3 Antibodies

          Invitrogen

          FOXP3 Monoclonal Antibody (PCH101), Biotin, eBioscience™

          Advanced Verification
          62 Published Figures
          62 References
          View all (149) FOXP3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite FOXP3 Monoclonal Antibody (PCH101), Biotin, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (62)
          • Advanced Verification (1)
          FOXP3 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FOXP3 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 64

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          FOXP3 Antibody (13-4776-82) in Flow

          Surface staining of normal human peripheral blood cells with Anti-Human CD4 FITC (Product # 11-0049-42) (left) and Anti-Human CD25 APC (Product # 17-0259-42) (right), followed by intracellular staining with 0.25 µg of Anti-Human Foxp3 Biotin or Rat IgG2a K Isotype Control Biotin (Product # 13-4321-82) followed by Streptavidin PE (Product # 12-4317-87) using the Foxp3/Transcription Factor Staining Buffer Set (Product # 00-5523-00). Cells in the lymphocyte gate were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Western Blot, Flow Cytometry (WB, Flow)
          FOXP3 Antibody in Western Blot, Flow Cytometry (WB, Flow)
          FOXP3 Antibody in Western Blot (WB)
          FOXP3 Antibody in Western Blot (WB)
          FOXP3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Immunohistochemistry (IHC)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody in Flow Cytometry (Flow)
          FOXP3 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          13-4776-82

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-Dependent
          View 1 publication 1 publication

          Immunohistochemistry (IHC)

          -
          View 15 publications 15 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-Dependent
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-Dependent
          -

          Flow Cytometry (Flow)

          0.25 µg/test
          View 44 publications 44 publications
          Product Specifications

          Species Reactivity

          Chimpanzee, Cynomolgus monkey, Human, Non-human primate, Rhesus monkey

          Published species

          Human

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), Biotin, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          PCH101

          Conjugate

          Biotin Biotin Biotin
          • Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 532 
          • Alexa Fluor 561
          • Alexa Fluor 660 
          • Alexa Fluor 700 
          • Alexa Fluor Plus 594 
          • Alexa Fluor Plus 647 
          • APC 
          • eFluor 450
          • eFluor 660
          • FITC
          • PE 
          • PE-Cyanine5
          • PE-Cyanine5.5
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-Cyanine5.5
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, do not freeze

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_763539

          Product Specific Information

          Description: eBioscience offers a panel of monoclonal antibodies to different epitopes of human Foxp3, providing useful tools for investigating the complete expression pattern of Foxp3 at the protein level, and discerning the precise subsets of Foxp3^+ cells.

          The PCH101 antibody reacts with the amino terminus of human foxp3 protein also known as FORKHEAD BOX P3, SCURFIN, and JM2; cross reactivity of this antibody to other proteins has not been determined. Foxp3, a 49-55 kDa protein, is a member of the forkhead/winged-helix family of transcriptional regulators, and was identified as the gene defective in 'scurfy' (sf) mice. Constitutive high expression of Foxp3 mRNA has been shown in CD4+CD25+ regulatory T cells (Treg cells), and ectopic expression of foxp3 in CD4+CD25- cells imparts a Treg phenotype in these cells.

          Intracellular staining of human peripheral blood mononuclear cells (PBMCs) with PCH101 antibody using the anti-human Foxp3 Staining Set and protocol reveals approximately 0.5-4% of lymphocytes staining, with the majority of staining occurring in the CD25^bright population. This is subject to donor variability.

          PCH101 cross-reacts with rhesus, chimpanzee and cynomolgus. We recommend the use of CD4 (OKT4, Product # 11-0048-42 , or RPA-T4, Product # 11-0049-42 , depending on the species) and CD25 (BC96, Product # 17-0259-42).

          Applications Reported: This PCH101 antibody has been reported for use in intracellular staining followed by flow cytometric analysis, western blotting, immunohistochemical staining of frozen and formalin-fixed paraffin embedded tissue sections.

          Applications Tested: This PCH101 antibody has been tested by intracellular flow cytometric analysis of normal human peripheral blood cells using the Foxp3/Transcription Factor Staining Buffer Set (Product # 00-5523) and protocol. Please see BestProtocols® Section (Staining intracellular Antigens for Flow Cytometry) for staining protocol (refer to Protocol B: One-step protocol for intracellular (nuclear) proteins). This antibody can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          FOXP3 (Forkhead box protein 3) is a member of the forkhead/winged-helix family of transcriptional regulators, highly conserved across mammals, and essential for normal immune homeostasis. FOXP3 is 381 amino acids long, stably and constitutively expressed at a high level in CD25 + CD4 positive regulatory T cells, a low level in CD4-positive/CD25-negative cells, and is absent in CD4-negative/CD8-positive T cells. FOXP3 may be a master regulatory gene, and a more specific marker of regulatory T cells. Defects in the gene encoding FOXP3 protein cause the scurfy phenotype in mice. In humans FOXP3 defects play a role in IPEX syndrome (immune dysfunction, polyendocrinopathy, enteropathy, X-linked syndrome), also known as X-linked autoimmunity-allergic dysregulation (XLAAD) syndrome. Transcript variants of FOXP3 encoding different isoforms have been identified. In human breast and colon cancer cells, expression of FOXP3 is regulated by p53 in response to the DNA damage.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: forkhead box P3; forkhead box P3 transcript variant; Forkhead box protein P3; FOXP3delta7; immune dysregulation, polyendocrinopathy, enteropathy, X-linked; immunodeficiency, polyendocrinopathy, enteropathy, X-linked; JM2; MGC141961; MGC141963; regulatory protein Foxp3; RP23-54C14.1; scurfin; unnamed protein product

          View more View less

          Gene Aliases: AIID; DIETER; IPEX; JM2; PIDX; XPID

          View more View less

          UniProt ID: (Cynomolgus monkey) Q6U8D7, (Human) Q9BZS1

          View more View less

          Entrez Gene ID: (Cynomolgus monkey) 102120882, (Human) 50943, (Rhesus monkey) 574303

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding transcription cofactor activity transcription corepressor activity protein binding histone acetyltransferase binding identical protein binding protein homodimerization activity histone deacetylase binding sequence-specific DNA binding NF-kappaB binding NFAT protein binding sequence-specific double-stranded DNA binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter response to reactive oxygen species negative regulation of cytokine production CD4-positive, CD25-positive, alpha-beta regulatory T cell lineage commitment regulation of T cell anergy negative regulation of T cell cytokine production chromatin remodeling regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter negative regulation of cell proliferation response to virus establishment of endothelial blood-brain barrier response to lipopolysaccharide negative regulation of interferon-gamma production negative regulation of interleukin-10 production negative regulation of interleukin-17 production negative regulation of interleukin-2 production negative regulation of interleukin-4 production positive regulation of CD4-positive, CD25-positive, alpha-beta regulatory T cell differentiation T cell activation negative regulation of T cell proliferation T cell homeostasis regulatory T cell differentiation positive regulation of regulatory T cell differentiation negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter negative regulation of activated T cell proliferation negative regulation of defense response to virus negative regulation of immune response response to rapamycin negative regulation of T-helper 17 cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-5d6d8bc979-gw77j:80/100.66.76.64:80.
          git-commit: e71199a02a6c1449b2ac9ded8316d5734e8aa2de
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.62.0-2026.11.45-1.0