Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • MHC Class I (H-2Kd) Antibodies

          Invitrogen

          MHC Class I (H-2Kd) Monoclonal Antibody (SF1-1.1.1), Biotin, eBioscience™

          9 Published Figures
          8 References
          View all (30) MHC Class I (H-2Kd) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite MHC Class I (H-2Kd) Monoclonal Antibody (SF1-1.1.1), Biotin, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (9)
          MHC Class I (H-2Kd) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          MHC Class I (H-2Kd) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 10

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MHC Class I (H-2Kd) Antibody (13-5957-82) in Flow

          Staining of BALB/c (left) and C57BL/6 (right) splenocytes with 0.125 µg of Mouse IgG2a K Isotype Control Biotin (Product # 13-4724-85) (blue histogram) or 0.125 µg of Anti-Mouse MHC Class I (H-2Kd) Biotin (purple histogram) followed by Streptavidin PE (Product # 12-4317-87). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MHC Class I (H-2Kd) Antibody in Flow Cytometry (Flow)
          MHC Class I (H-2Kd) Antibody in Flow Cytometry (Flow)
          MHC Class I (H-2Kd) Antibody in Flow Cytometry (Flow)
          MHC Class I (H-2Kd) Antibody in Flow Cytometry (Flow)
          MHC Class I (H-2Kd) Antibody in Flow Cytometry (Flow)
          MHC Class I (H-2Kd) Antibody in Flow Cytometry (Flow)
          MHC Class I (H-2Kd) Antibody in Flow Cytometry (Flow)
          MHC Class I (H-2Kd) Antibody in Flow Cytometry (Flow)
          MHC Class I (H-2Kd) Antibody in Flow Cytometry (Flow)
          MHC Class I (H-2Kd) Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          13-5957-82

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          0.25 µg/test
          View 8 publications 8 publications
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Mouse

          Host/Isotype

          Mouse / IgG2a, kappa

          Recommended Isotype Control

          Mouse IgG2a kappa Isotype Control (eBM2a), Biotin, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          SF1-1.1.1

          Conjugate

          Biotin Biotin Biotin
          • APC 
          • APC-eFluor 780
          • eFluor 450
          • Functional Grade
          • PE 
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, do not freeze

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_1582234

          Product Specific Information

          Description: This SF1-1.1.1 monoclonal antibody reacts with the H-2Kd MHC class I alloantigen. H-2Kd is involved in antigen presentation to T cells expressing CD3/TCR and CD8. Reactivity to other haplotypes (e.g. b, j, k, p, q, s, and v) has not been observed.

          Applications Reported: This SF1-1.1.1 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This SF1-1.1.1 antibody has been tested by flow cytometric analysis of Balb/c splenocytes. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          This protein is an MHC class I alloantigen, which is involved in antigen presentation to T cells expressing CD3/TCR and CD8.

          HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: class I major histocompatibility antigen H-2Kd; H-2 class I histocomatibility antigen, K-k alpha chain, topological domain; H-2 class I histocompatibility antigen, K-B alpha chain; H-2 class I histocompatibility antigen, K-D alpha chain; H-2 class I histocompatibility antigen, K-K alpha chain; H-2 class I histocompatibility antigen, K-Q alpha chain; H-2K(B); H-2K(K); H-2K(Q); H2-K; histocompatibility 2, K region, promoter 1; MGC184092; MGC7052; MHC class I alpha chain; MHC class I antigen; MHC class I H2 antigen; MHC class I H2-K-b-alpha-2 cell surface glycoprotein; MHC class I heavy chain H2-K; MHC class I heavy chain heavy chain; MHC class I heavy chain maturation peptide; MHC class II antigen D; MHC class II antigen K; MHC H-2K integral membrane protein; MHC H-2K transplantation antigen; MHC H-2K-kml mRNA; MHC H2-K antigen; MHC H2-K transplantation antigen; MHC I; MHCI; ORF; transmembrane protein; unnamed protein product

          View more View less

          Gene Aliases: H-2K; H-2K(d); H2-D1; H2-K; K-f

          View more View less

          UniProt ID: (Mouse) P01901

          View more View less

          Entrez Gene ID: (Mouse) 14972

          View more View less

          Function(s)
          receptor binding protein binding beta-2-microglobulin binding MHC class I protein binding peptide antigen binding T cell receptor binding CD8 receptor binding identical protein binding protein homodimerization activity natural killer cell lectin-like receptor binding TAP binding TAP1 binding TAP2 binding receptor agonist activity chaperone binding TAP complex binding 14-3-3 protein binding
          Process(es)
          positive regulation of T cell mediated cytotoxicity antigen processing and presentation of endogenous peptide antigen via MHC class Ib antigen processing and presentation of endogenous peptide antigen via MHC class I via ER pathway, TAP-dependent antigen processing and presentation of endogenous peptide antigen via MHC class I via ER pathway, TAP-independent immune response signal transduction negative regulation of neuron projection development antigen processing and presentation antigen processing and presentation of exogenous peptide antigen via MHC class I defense response to bacterium inner ear development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-dpq8n:80/100.66.76.11:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0