Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • HuA/HuB/HuC/HuD Antibodies

          Proteintech

          HuA/B/C/D Polyclonal Antibody

          5 References

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite HuA/B/C/D Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (7)
          HuA/B/C/D Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          HuA/B/C/D Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          HuA/B/C/D Antibody (13032-1-AP) in IHC (P)

          Immunohistochemistry of paraffin-embedded human brain tissue slide using 13032-1-AP ( HuA/B/C/D antibody at dilution of 1:200 (under 40x lens). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          HuA/B/C/D Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HuA/B/C/D Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HuA/B/C/D Antibody in Western Blot (WB)
          HuA/B/C/D Antibody in Western Blot (WB)
          HuA/B/C/D Antibody in Western Blot (WB)
          HuA/B/C/D Antibody in Western Blot (WB)
          HuA/B/C/D Antibody in Immunoprecipitation (IP)
          HuA/B/C/D Polyclonal Antibody

          Product Details

          13032-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:4,000
          View 1 publication 1 publication

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          -
          View 2 publications 2 publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          HuA/B/C/D Fusion Protein Ag3678 (1-366 aa encoded by BC036071)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.17 mg/mL

          Amount

          25.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MVMIISTME PQVSNGPTSN TSNGPSSNNR NCPSPMQTGA TTDDSKTNLI VNYLPQNMTQ EEFRSLFGSI GEIESCKLVR DKITGQSLGY GFVNYIDPKD AEKAINTLNG LRLQTKTIKV SYARPSSASI RDANLYVSGL PKTMTQKELE QLFSQYGRII TSRILVGQVT GVSRGVGFIR FDKRIEAEEA IKGLNGQKPS GATEPITVKF ANNPSQKSSQ ALLSQLYQSP NRRYPGPLHH QAQRFRLDNL LNMAYGVKRF SPITIDGMTS LVGMNIPGHT GTGWCIFVYN LSPDSDESVL WQLFGPFGAV NNVKVIRDFN TNKCKGFGFV TMTNYDEAAM AITSLNGYRL GDRVLQVSFK TNKAHKS (1-366 aa encoded by BC036071)

          Target Information

          Embryonic lethal, abnormal vision, Drosophila-like 4 (ELAVL4) is a human homologue of the Drosophila gene ELAV, which is implicated in neuronal differentiation and maintenance. The ELAVL proteins are mRNA binding proteins and all members of the family contain three RNA-recognition motifs. ELAVL-proteins also interact with inflammation-associated factors. The protein may be implicated in neurodegeneration through these complex mechanisms.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ELAV (embryonic lethal, abnormal vision, Drosophila)-like 1 (Hu antigen R); ELAV (embryonic lethal, abnormal vision, Drosophila)-like 2 (Hu antigen B); ELAV (embryonic lethal, abnormal vision, Drosophila)-like 3 (Hu antigen C); ELAV (embryonic lethal, abnormal vision, Drosophila)-like 4 (Hu antigen D); ELAV like 1; ELAV like neuron-specific RNA binding protein 2; ELAV like neuron-specific RNA binding protein 3; ELAV like neuron-specific RNA binding protein 4; Elav-like generic protein; ELAV-like neuronal protein 1; ELAV-like protein 1; ELAV-like protein 2; ELAV-like protein 3; ELAV-like protein 4; embryonic lethal, abnormal vision, drosophila, homolog-like 1; embryonic lethal, abnormal vision-like 2; Hu antigen C; Hu antigen D; Hu antigen R; HU-antigen A; HU-antigen B; HU-antigen C; HU-antigen D; Hu-antigen R; HuB; HuC; HuD; Human antigen R; HuR; MelG; Nervous system-specific RNA-binding protein Hel-N1; Nervous system-specific RNA-binding protein Mel-N1; Paraneoplastic cerebellar degeneration-associated antigen; paraneoplastic encephalomyelitis antigen; Paraneoplastic encephalomyelitis antigen HuD; Paraneoplastic limbic encephalitis antigen 21; RNA binding protein HuB; RNA-binding protein HUD3; unnamed protein product

          View more View less

          Gene Aliases: 2410055N02Rik; 2600009P04Rik; Elav; ELAV1; Elav2; ELAVL1; ELAVL2; ELAVL3; ELAVL4; Elra; HEL-N1; HELN1; Hua; HUB; HUC; HUCL; HUD; HUR; mel-N1; MelG; mHuC; PLE21; PNEM; r-HuD; RGD1561943; W91709

          View more View less

          UniProt ID: (Human) Q15717, (Human) Q12926, (Human) Q14576, (Human) P26378, (Mouse) P70372, (Rat) B5DF91, (Mouse) Q60899, (Mouse) Q60900, (Mouse) Q61701, (Rat) O09032

          View more View less

          Entrez Gene ID: (Human) 1994, (Human) 1993, (Human) 1995, (Human) 1996, (Mouse) 15568, (Rat) 363854, (Mouse) 15569, (Rat) 286973, (Mouse) 15571, (Rat) 282824, (Mouse) 15572, (Rat) 432358

          View more View less

          Function(s)
          nucleic acid binding RNA binding double-stranded RNA binding mRNA binding mRNA 3'-UTR binding protein binding protein kinase binding miRNA binding mRNA 3'-UTR AU-rich region binding protein homodimerization activity lncRNA binding poly(A) binding pre-mRNA intronic pyrimidine-rich binding nucleotide binding AU-rich element binding poly(A) RNA binding translation initiation factor
          Process(es)
          lncRNA-mediated post-transcriptional gene silencing protein import into nucleus cell proliferation response to glucose positive regulation of autophagy posttranscriptional gene silencing positive regulation of superoxide anion generation regulation of mRNA stability positive regulation of translation positive regulation of autophagosome size mRNA stabilization protein homooligomerization negative regulation of gene silencing by miRNA mRNA destabilization 3'-UTR-mediated mRNA stabilization regulation of stem cell population maintenance regulation of transcription, DNA-templated nervous system development cell differentiation RNA processing mRNA processing learning locomotory behavior RNA splicing cerebral cortex neuron differentiation dendrite morphogenesis positive regulation of 3'-UTR-mediated mRNA stabilization regulation of translation multicellular organism development neuron differentiation positive regulation of dendrite development aging associative learning regeneration response to endoplasmic reticulum stress response to cocaine cellular response to nerve growth factor stimulus
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline