Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • MTUS1 Antibodies

          Proteintech

          MTUS1 Polyclonal Antibody

          7 References
          View all (13) MTUS1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite MTUS1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (10)
          MTUS1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          MTUS1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 10

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MTUS1 Antibody (13436-1-AP) in ICC/IF

          Immunofluorescent analysis of ( -20°C Ethanol ) fixed PC-3 cells using 13436-1-AP (MTUS1 antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MTUS1 Antibody in Immunocytochemistry (ICC/IF)
          MTUS1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          MTUS1 Antibody in Western Blot (WB)
          MTUS1 Antibody in Western Blot (WB)
          MTUS1 Antibody in Western Blot (WB)
          MTUS1 Antibody in Western Blot (WB)
          MTUS1 Antibody in Western Blot (WB)
          MTUS1 Antibody in Western Blot (WB)
          MTUS1 Antibody in Western Blot (WB)
          MTUS1 Antibody in Immunoprecipitation (IP)
          MTUS1 Polyclonal Antibody

          Product Details

          13436-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 4 publications 4 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:600
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          View 2 publications 2 publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          MTUS1 Fusion Protein Ag4242 (196-436 aa encoded by BC033842)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.13 mg/mL

          Amount

          19.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          The antibody can recognize all isoforms except isoform 5 (85-90 kDa). The antibody tested HomoDimer isoforms (80 kDa/110-120/280 kDa) in Jurkat cell.

          Immunogen sequence: MQLQEQFDN LNAAHETSKL EIEASHSEKL ELLTKAYEAS LSEIKKGHEI EKKSLEDLLS EKQESLEKQI NDLKSENDAL NEKLKSEEQK RRAREKANLK NPQIMYLEQE LESLKAVLEI KNEKLHQQDI KLMKMEKLVD NNTALVDKLK RFQQENEELK ARMDKHMAIS RQLSTEQAVL QESLEKESKV NKRLSMENEE LLWKLHNGDL CSPKRSPTSS AIPLQSPRNS GSFPSPSISP R (196-436 aa encoded by BC033842)

          Target Information

          The protein belongs to the FKBP-type peptidyl-prolyl cis/trans isomerase family. It is located in endoplasmic reticulum and acts as molecular chaperones.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: angiotensin II AT2 receptor-interacting protein 1; Angiotensin-II type 2 receptor-interacting protein; AT2 receptor-binding protein; AT2 receptor-interacting protein; AT2 receptor-interacting protein 1; AT2R binding protein; ATIP1; Coiled-coiled tumor suppressor gene 1 protein; coiled-coiled tumor supressor gene 1; DKFZp586D1519; DKFZp686F20243; erythroid differentiation-related; FLJ14295; growth factor inhibitor; HSNEP; microtubule associated tumor suppressor 1; Microtubule-associated tumor suppressor 1; Microtubule-associated tumor suppressor 1 homolog; Mitochondrial tumor suppressor 1; Mitochondrial tumor suppressor 1 homolog; mitochondrial tumor suppressor gene 1; transcription factor MTSG1; unnamed protein product

          View more View less

          Gene Aliases: AI481402; ATBP; ATBP135; ATIP; Atip1; ATIP3; Atip3b; ATIP4; B430010I23Rik; B430305I03Rik; C85752; Cctsg1; Cctsg1-440; GK1; ICIS; KIAA1288; MD44; mKIAA1288; MP44; MTSG1; MTUS1

          View more View less

          UniProt ID: (Human) Q9ULD2, (Mouse) Q5HZI1, (Rat) Q6IMY1

          View more View less

          Entrez Gene ID: (Human) 57509, (Mouse) 102103, (Rat) 306487

          View more View less

          Function(s)
          protein binding microtubule binding molecular_function
          Process(es)
          regulation of macrophage chemotaxis cell cycle cellular response to peptide hormone stimulus
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0