Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • gamma Catenin Antibodies

          Invitrogen

          gamma Catenin Monoclonal Antibody (PG-11E4)

          Advanced Verification
          17 Published Figures
          34 References
          View all (42) gamma Catenin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite gamma Catenin Monoclonal Antibody (PG-11E4)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (5)
          • Published Figures (17)
          • Advanced Verification (2)
          gamma Catenin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          gamma Catenin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 24

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          gamma Catenin Antibody (13-8500) in ICC/IF

          Immunofluorescence analysis of Gamma-Catenin was performed using 70% confluent log phase CACO2 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 1% BSA for 1 hour at room temperature. The cells were labeled with Gamma-Catenin / Plakoglobin (PG-11E4) Mouse Monoclonal Antibody (Product # 13-8500) at 1:250 dilution in0.1%... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          gamma Catenin Antibody in Immunocytochemistry (ICC/IF)
          gamma Catenin Antibody in Western Blot (WB)
          gamma Catenin Antibody in Western Blot (WB)
          gamma Catenin Antibody in Western Blot (WB)
          gamma Catenin Antibody in Western Blot (WB)
          gamma Catenin Antibody in Immunocytochemistry (ICC/IF)
          gamma Catenin Antibody in Immunocytochemistry (ICC/IF)
          gamma Catenin Antibody in Immunocytochemistry (ICC/IF)
          gamma Catenin Antibody in Immunocytochemistry (ICC/IF)
          gamma Catenin Antibody in Immunocytochemistry (ICC/IF)
          gamma Catenin Antibody in Immunocytochemistry (ICC/IF)
          gamma Catenin Antibody in Immunocytochemistry (ICC/IF)
          gamma Catenin Antibody in Immunocytochemistry (ICC/IF)
          gamma Catenin Antibody in Immunocytochemistry (ICC/IF)
          gamma Catenin Antibody in Immunohistochemistry (IHC)
          gamma Catenin Antibody in Western Blot (WB)
          gamma Catenin Antibody in Western Blot (WB)
          gamma Catenin Antibody in Western Blot, Immunoprecipitation (WB, IP)
          gamma Catenin Antibody in Western Blot (WB)
          gamma Catenin Antibody in Western Blot, Immunoprecipitation (WB, IP)
          gamma Catenin Antibody in Western Blot (WB)
          gamma Catenin Antibody in Immunoprecipitation (IP)
          gamma Catenin Antibody
          gamma Catenin Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          gamma Catenin Monoclonal Antibody (PG-11E4)

          Product Details

          13-8500

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:250
          View 17 publications 17 publications

          Immunohistochemistry (IHC)

          Assay-dependent
          View 4 publications 4 publications

          Immunocytochemistry (ICC/IF)

          1:250
          View 7 publications 7 publications

          Immunoprecipitation (IP)

          Assay-dependent
          View 3 publications 3 publications

          Miscellaneous PubMed (Misc)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          PG-11E4

          Immunogen

          Fusion protein consisting of the maltose binding protein fused to full length human plakoglobin protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533040

          Target Information

          JUP is a major cytoplasmic protein which is the only known constituent common to sub membranous plaques of both desmosomes and intermediate junctions. This protein forms distinct complexes with cadherins and desmosomal cadherins and is a member of the catenin family since it contains a distinct repeating amino acid motif called the armadillo repeat.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: catenin (cadherin-associated protein), gamma 80kDa; desmoplakin III; desmoplakin-3; desmosomal protein 3; gamma-catenin; OTTHUMP00000164735; unnamed protein product

          View more View less

          Gene Aliases: CTNNG; D930025P04Rik; DP3; DPIII; PDGB; PG; PKGB

          View more View less

          UniProt ID: (Human) P14923, (Mouse) Q02257

          View more View less

          Entrez Gene ID: (Human) 3728, (Mouse) 16480

          View more View less

          Function(s)
          transcription coactivator activity structural molecule activity protein binding ligand-dependent nuclear receptor binding protein phosphatase binding alpha-catenin binding cadherin binding cell adhesion molecule binding cell adhesive protein binding involved in bundle of His cell-Purkinje myocyte communication microtubule cortical anchor activity protein kinase binding macromolecular complex binding protein tyrosine kinase binding
          Process(es)
          positive regulation of cell-matrix adhesion desmosome assembly cell adhesion cell migration regulation of cell proliferation positive regulation of protein import into nucleus negative regulation of blood vessel endothelial cell migration positive regulation of angiogenesis positive regulation of axon extension positive regulation of transcription from RNA polymerase II promoter detection of mechanical stimulus canonical Wnt signaling pathway endothelial cell-cell adhesion cellular response to indole-3-methanol protein localization to plasma membrane bundle of His cell-Purkinje myocyte adhesion involved in cell communication regulation of heart rate by cardiac conduction positive regulation of canonical Wnt signaling pathway cell-cell adhesion regulation of ventricular cardiac muscle cell action potential positive regulation of cellular protein localization skin development cell-cell junction organization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7bf6cff7cf-x9r6b:80/100.66.78.146:80.
          git-commit: cd21911f92a61daaa7859949453b3286702c44a2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.7.0-2026.11.47-1.0