Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD86 Antibodies

          Invitrogen

          CD86 (B7-2) Monoclonal Antibody (GL1), eBioscience™

          98 Published Figures
          159 References
          View all (225) CD86 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD86 (B7-2) Monoclonal Antibody (GL1), eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (98)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 99

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD86 (B7-2) Antibody (14-0862-85) in Flow

          Staining of 3-day LPS-stimulated BALB/c splenocytes with 0.25 µg of Rat IgG2a kappa Isotype Control Purified (Product # 14-4321-82) (open histogram) or 0.25 µg of Purified Anti-Mouse CD86 (B7-2) (filled histogram) followed by Anti-Rat IgG FITC (Product # 11-4811-85). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Immunocytochemistry (ICC/IF)
          CD86 (B7-2) Antibody in Immunocytochemistry (ICC/IF)
          CD86 (B7-2) Antibody in Immunohistochemistry (IHC)
          CD86 (B7-2) Antibody in Immunohistochemistry (IHC)
          CD86 (B7-2) Antibody in Immunohistochemistry (IHC)
          CD86 (B7-2) Antibody in Immunohistochemistry (IHC)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)
          CD86 (B7-2) Antibody in Flow Cytometry (Flow)

          Product Details

          14-0862-85

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 11 publications 11 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-Dependent
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          -
          View 5 publications 5 publications

          Flow Cytometry (Flow)

          0.5 µg/test
          View 136 publications 136 publications

          ELISA (ELISA)

          -
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          Assay-Dependent
          View 1 publication 1 publication

          Neutralization (Neu)

          Assay-Dependent
          View 2 publications 2 publications

          Functional Assay (Functional)

          Assay-Dependent
          -
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rat / IgG2a, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          GL1

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 700 
          • APC 
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 737
          • Brilliant Violet 480
          • Brilliant Violet 605
          • Brilliant Violet 786
          • eFluor 450
          • FITC
          • PE 
          • PE-Cyanine5
          • PE-Cyanine7
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Super Bright 702
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          500 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_467369

          Product Specific Information

          Description: The GL1 monoclonal antibody reacts with mouse CD86, an ~80 kDa surface receptor also known as B7-2. CD86 and CD80 are members of the B7 family of costimulatory molecules. CD86 is expressed at low level on B cells, macrophages, and dendritic cells and is upregulated on B cells through a variety of surface stimuli including the BCR complex, CD40 and some cytokine receptors. CD86 is also expressed by activated mouse T cells and thioglycolate-elicited peritoneal cells. In addition to CD80 (B7-1), CD86 is a counter-receptor for the T cell surface molecules CD28 and CD152 (CTLA-4). This interaction plays a critical role in T-B crosstalk, T cell costimulation, autoantibody production and Th2-mediated Ig production. The kinetics of upregulation of CD86 upon stimulation, supports its major contribution during the primary phase of an immune response.

          Applications Reported: This GL1 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunohistology staining of frozen tissue sections. It has also been reported in blocking of CD86 in functional studies. (Please use Functional Grade purified GL1, Product # 16-0862, in functional assays).

          Applications Tested: The GL1 antibody has been tested by flow cytometric analysis of resting and activated mouse splenocytes. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          CD86, along with CD80, is a member of the B7 family of costimulatory molecules and plays a crucial role in T cell activation and immune response regulation. CD86 is expressed at low levels on B cells, macrophages, and dendritic cells, and its expression is upregulated on B cells through various stimuli, including the BCR complex, CD40, and certain cytokine receptors. As a type I membrane protein and member of the immunoglobulin superfamily, CD86 serves as a ligand for the T cell surface proteins CD28 and CTLA-4 (CD152). The interaction between CD86 and CD28 provides a costimulatory signal essential for T cell activation during antigen presentation, while binding with CTLA-4 negatively regulates T cell activation, diminishing the immune response. This interaction is critical for T-B cell crosstalk, T cell costimulation, autoantibody production, and Th2-mediated Ig production. The kinetics of CD86 upregulation upon stimulation suggest its significant contribution during the primary phase of an immune response. CD86 and CD80 have distinct roles in T helper cell differentiation, and insufficient co-stimulation involving these molecules can induce tolerance. Alternative splicing of CD86 results in two transcript variants encoding different isoforms, with additional variants described but not fully sequenced. Dysfunction in CD86 is associated with diseases such as gallbladder squamous cell carcinoma and myocarditis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Activation B7-2 antigen; B lymphocyte activation antigen B72; CD28 antigen ligand 2; CD86; early T cell costimulatory molecule-1; Early T-cell co-stimulatory molecule 1; Early T-cell costimulatory molecule 1; ETC-1; MGC34413; T-lymphocyte activation antigen CD86

          View more View less

          Gene Aliases: B7; B7-2; B7.2; B70; Cd28l2; Cd86; CLS1; ETC-1; Ly-58; Ly58; MB7; MB7-2; TS/A-2

          View more View less

          UniProt ID: (Mouse) P42082

          View more View less

          Entrez Gene ID: (Mouse) 12524

          View more View less

          Function(s)
          receptor binding immunoglobulin receptor superfamily
          Process(es)
          toll-like receptor signaling pathway immune system process negative regulation of T cell anergy positive regulation of T cell proliferation positive regulation of activated T cell proliferation B cell activation defense response to virus cellular response to lipopolysaccharide
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline