Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 新產品
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • CD90.1 (Thy1.1) Antibodies

          Invitrogen

          CD90.1 (Thy-1.1) Monoclonal Antibody (HIS51), eBioscience™

          50 Published Figures
          91 References
          View all (18) CD90.1 (Thy1.1) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD90.1 (Thy-1.1) Monoclonal Antibody (HIS51), eBioscience™

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (50)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 51

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD90.1 (Thy-1.1) Antibody (14-0900-81) in Flow

          Surface staining of rat splenocytes with Anti-Mouse/Rat CD90.1 (Thy-1.1) FITC (left) and PE (right). Appropriate isotype controls were used (open histogram). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Immunocytochemistry (ICC/IF)
          CD90.1 (Thy-1.1) Antibody in Immunocytochemistry (ICC/IF)
          CD90.1 (Thy-1.1) Antibody in Immunocytochemistry (ICC/IF)
          CD90.1 (Thy-1.1) Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          CD90.1 (Thy-1.1) Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          CD90.1 (Thy-1.1) Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
          CD90.1 (Thy-1.1) Antibody in Immunohistochemistry (IHC)
          CD90.1 (Thy-1.1) Antibody in Immunohistochemistry (IHC)
          CD90.1 (Thy-1.1) Antibody in Immunohistochemistry (IHC)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)
          CD90.1 (Thy-1.1) Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          14-0900-81

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 6 publications 6 publications

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-Dependent
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 4 publications 4 publications

          Flow Cytometry (Flow)

          0.125 µg/test
          View 78 publications 78 publications

          Functional Assay (Functional)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG2a, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          HIS51

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • APC 
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 496
          • Brilliant UV 615
          • Brilliant Violet 786
          • eFluor 450
          • FITC
          • PE 
          • PE-Cyanine5
          • PE-Cyanine7
          • PerCP-Cyanine5.5
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_467373

          Product Specific Information

          Description: The HIS51 monoclonal antibody reacts with rat CD90 and cross-reacts with mouse CD90.1, a GPI-linked membrane molecule. In the rat, CD90 is expressed by hematopoietic stem cells, immature B cells, thymocytes, recent thymic emigrants, neurons, inflammed endothelia and other cell types. In the CD90.1-expressing mouse strains, PL and AKR, CD90 is expressed by early hematopoietic cells in the bone marrow, thymocytes and mature T cells.

          Applications Reported: The HIS51 antibody has been reported for use in flow cytometric analysis, and immunohistochemical staining of frozen tissue sections.

          Applications Tested: The HIS51 antibody has been tested by flow cytometric analysis of rat thymocyte and splenocyte suspensions. This can be used at less than or equal to 0.125 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          CD90 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. It is expressed in murine T cells, thymocytes, neural cells, Kupffer's cells and fibroblasts. Thy 1 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CD90; cell surface protein; FLJ33325; thy-1 antigen; thy-1 glycoprotein; Thy-1 protein; Thy-1 protein precursor; thymus cell antigen 1, theta; Thymus cell surface antigen; unnamed protein product

          View more View less

          Gene Aliases: CD7; CD90; T25; Thy-1; Thy-1.2; Thy1.1; Thy1.2

          View more View less

          UniProt ID: (Rat) P01830, (Mouse) P01831

          View more View less

          Entrez Gene ID: (Rat) 24832, (Mouse) 21838

          View more View less

          Function(s)
          GTPase activator activity integrin binding protein binding enzyme binding protein kinase binding GPI anchor binding
          Process(es)
          angiogenesis regulation of cell-matrix adhesion regulation of immune system process positive regulation of cellular extravasation negative regulation of protein kinase activity cytoskeleton organization cell adhesion cell surface receptor signaling pathway integrin-mediated signaling pathway cell-cell signaling regulation of cell migration negative regulation of cell migration heterotypic cell-cell adhesion negative regulation of apoptotic process receptor clustering positive regulation of GTPase activity mast cell activation positive regulation of cell adhesion retinal cone cell development protein autophosphorylation focal adhesion assembly negative regulation of fibroblast proliferation response to axon injury positive regulation of peptidyl-tyrosine phosphorylation negative regulation of axonogenesis T cell receptor signaling pathway negative regulation of T cell receptor signaling pathway positive regulation of T cell activation positive regulation of release of sequestered calcium ion into cytosol positive regulation of focal adhesion assembly negative regulation of neuron projection regeneration cell-cell adhesion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-79565875f6-kzhm8:80/100.66.77.38:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline