Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • c-Kit (CD117) Antibodies

          Invitrogen

          CD117 (c-Kit) Monoclonal Antibody (ACK2), eBioscience™

          48 Published Figures
          85 References
          View all (221) c-Kit (CD117) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD117 (c-Kit) Monoclonal Antibody (ACK2), eBioscience™

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (48)
          CD117 (c-Kit) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CD117 (c-Kit) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 49

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD117 (c-Kit) Antibody (14-1172-81) in WB

          Western blot was performed using Anti-CD117 (c-Kit) Monoclonal Antibody (ACK2), eBioscience™ (Product # 14-1172-82) and a 55kDa band corresponding to Mast/stem cell growth factor receptor Kit was observed across tissues tested. Whole cell extracts (30 µg lysate) of Mouse Lung (Lane 1), Mouse Bone Marrow (Lane 2), Rat Bone Marrow (Lane 3), Mouse Kidney (Lane 4), Rat Kidney (Lane 5), Rat Lung (Lane 6), Mouse Spleen (Lane 7), Rat Spleen (Lane 8), Mouse Thymus (Lane 9) were electrophoresed using NuPAGE™ 4-12% Bis-Tris Protein Gel (Product # NP... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD117 (c-Kit) Antibody in Western Blot (WB)
          CD117 (c-Kit) Antibody in Immunocytochemistry (ICC/IF)
          CD117 (c-Kit) Antibody in Immunocytochemistry (ICC/IF)
          CD117 (c-Kit) Antibody in Immunocytochemistry (ICC/IF)
          CD117 (c-Kit) Antibody in Immunocytochemistry (ICC/IF)
          CD117 (c-Kit) Antibody in Immunocytochemistry (ICC/IF)
          CD117 (c-Kit) Antibody in Immunocytochemistry (ICC/IF)
          CD117 (c-Kit) Antibody in Immunocytochemistry (ICC/IF)
          CD117 (c-Kit) Antibody in Immunocytochemistry (ICC/IF)
          CD117 (c-Kit) Antibody in Immunocytochemistry (ICC/IF)
          CD117 (c-Kit) Antibody in Immunocytochemistry (ICC/IF)
          CD117 (c-Kit) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD117 (c-Kit) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD117 (c-Kit) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD117 (c-Kit) Antibody in Immunohistochemistry (IHC)
          CD117 (c-Kit) Antibody in Immunohistochemistry (IHC)
          CD117 (c-Kit) Antibody in Immunohistochemistry (IHC)
          CD117 (c-Kit) Antibody in Immunohistochemistry (IHC)
          CD117 (c-Kit) Antibody in Immunohistochemistry (IHC)
          CD117 (c-Kit) Antibody in Immunohistochemistry (IHC)
          CD117 (c-Kit) Antibody in Immunohistochemistry (IHC)
          CD117 (c-Kit) Antibody in Immunohistochemistry (IHC)
          CD117 (c-Kit) Antibody in Immunohistochemistry (IHC)
          CD117 (c-Kit) Antibody in Immunohistochemistry (IHC)
          CD117 (c-Kit) Antibody in Immunohistochemistry (IHC)
          CD117 (c-Kit) Antibody in Immunohistochemistry (IHC)
          CD117 (c-Kit) Antibody in Immunohistochemistry (IHC)
          CD117 (c-Kit) Antibody in Western Blot (WB)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)
          CD117 (c-Kit) Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          14-1172-81

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 4 publications 4 publications

          Immunohistochemistry (IHC)

          -
          View 26 publications 26 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-Dependent
          -

          Immunocytochemistry (ICC/IF)

          -
          View 14 publications 14 publications

          Flow Cytometry (Flow)

          0.125 µg/test
          View 35 publications 35 publications

          Immunoprecipitation (IP)

          Assay-Dependent
          -

          Neutralization (Neu)

          -
          View 1 publication 1 publication

          Functional Assay (Functional)

          Assay-Dependent
          View 2 publications 2 publications

          Inhibition Assays (Inhibition)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Chicken, Fish, Guinea pig, Human, Mouse, Rat

          Host/Isotype

          Rat / IgG2b, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          ACK2

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 700 
          • APC 
          • APC-eFluor 780
          • Functional Grade
          • PE 
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 702
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_467435

          Product Specific Information

          Description: The ACK2 monoclonal antibody reacts with mouse CD117, also known as c-Kit receptor, Steel factor receptor and stem cell factor receptor. A member of the tyrosine kinase receptor family, this 145 kDa molecule is expressed by a majority of hematopoietic progenitor cells characterized in the mouse bone marrow as a small subset of cells positive for Sca-1 and Thy1 (Thy1^lo) and negative for lineage markers. The interaction of the mouse c-kit receptor and steel factor promotes the proliferation and differentiation of hematopoietic progenitor cells. CD117 is also expressed by mast cells and plays a role in signaling and activation of these cells. ACK2 has been reported to be a blocking antibody.

          Applications Reported: The ACK2 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, immunoblotting (WB), and immunohistochemical staining of frozen tissue sections. It has also been reported for use in functional assays. (Please use Functional Grade purified ACK2, Product # 16-1172, in functional assays.) (Fluorochrome-conjugated ACK2 is recommended for use in flow cytometric analysis).

          Applications Tested: The ACK2 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.125 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          KIT (c-KIT) is a proto-oncogene and a type 3 transmembrane receptor for MGF (mast cell growth factor, also known as stem cell factor). KIT was first identified as the cellular homolog of the feline sarcoma viral oncogene v-kit. KIT together with its ligand regulates growth and activation of a variety of hemopoietic and non-hemopoietic cells. Mutations in KIT are associated with gastrointestinal stromal tumors, mast cell disease, acute myelogenous leukemia, and piebaldism. Recently, deregulation of the KIT receptor TK by the prevalent activation loop mutation D816V has served as a focal point in therapeutic strategies aimed at curbing neoplastic mast cell growth. c-Kit is expressed in hematopoietic stem cells, germ cells, mast cells and gastrointestinal tract cajal cells. Upon binding of its ligand stem cell factor (SCF), c-kit dimerizes, resulting in receptor activation and autophosphorylation of various tyrosine residues including tyrosine 703 located on the cytoplasmic domain of the receptor. This modification allows docking of Grb2 and activation of the Ras/ERK signaling pathway. SCF/c-kit can activate multiple downstream signaling pathways including PI3K, PLC-gamma and JAK/STAT. c-kit receptor activation is essential for hematopoiesis, stem cell maintenance and gametogenesis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: belly-spot; c-kit proto-oncogene protein; c-Kit protooncogene; c-kit receptor tyrosine kinase; ckit; dominant spotting; Dominant white spotting; Kit oncogene; p145 c-kit; piebald trait protein; protein kinase; proto-oncogene c-Kit; proto-oncogene tyrosine-protein kinase Kit; SCFR; similar to v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene; spotted sterile male; Steel Factor Receptor; tyrosine kinase; tyrosine-protein kinase Kit; unnamed protein product; v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog; v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein

          View more View less

          Gene Aliases: Bs; C-Kit; CD117; Fdc; Gsfsco1; Gsfsco5; Gsfsow3; MASTC; PBT; SCFR; SCO1; SCO5; SOW3; Ssm; Tr-kit; W

          View more View less

          UniProt ID: (Human) P10721, (Mouse) P05532

          View more View less

          Entrez Gene ID: (Human) 3815, (Rat) 64030, (Mouse) 16590

          View more View less

          Function(s)
          protease binding protein kinase activity protein tyrosine kinase activity transmembrane receptor protein tyrosine kinase activity stem cell factor receptor activity protein binding ATP binding growth factor binding cytokine binding SH2 domain binding protein homodimerization activity nucleotide binding enzyme binding metal ion binding
          Process(es)
          ovarian follicle development hematopoietic progenitor cell differentiation myeloid progenitor cell differentiation lymphoid progenitor cell differentiation immature B cell differentiation mast cell chemotaxis myeloid leukocyte differentiation positive regulation of cytokine production involved in immune response positive regulation of dendritic cell cytokine production protein phosphorylation glycosphingolipid metabolic process chemotaxis response to stress inflammatory response transmembrane receptor protein tyrosine kinase signaling pathway spermatogenesis spermatid development positive regulation of cell proliferation germ cell migration regulation of cell shape gonad development visual learning male gonad development response to radiation positive regulation of gene expression cell migration peptidyl-tyrosine phosphorylation cytokine-mediated signaling pathway lamellipodium assembly actin cytoskeleton organization hemopoiesis B cell differentiation T cell differentiation erythrocyte differentiation melanocyte differentiation positive regulation of cell migration positive regulation of pseudopodium assembly positive regulation of mast cell cytokine production somatic stem cell population maintenance embryonic hemopoiesis ectopic germ cell programmed cell death organ growth intracellular signal transduction hematopoietic stem cell migration megakaryocyte development Fc receptor signaling pathway Kit signaling pathway erythropoietin-mediated signaling pathway positive regulation of tyrosine phosphorylation of STAT protein negative regulation of programmed cell death mast cell degranulation positive regulation of MAP kinase activity positive regulation of MAPK cascade pigmentation tongue development leukocyte activation positive regulation of Notch signaling pathway positive regulation of JAK-STAT cascade response to cadmium ion protein autophosphorylation developmental pigmentation positive regulation of long-term neuronal synaptic plasticity digestive tract development stem cell differentiation epithelial cell proliferation regulation of developmental process detection of mechanical stimulus involved in sensory perception of sound cell chemotaxis mast cell differentiation response to growth factor melanocyte migration melanocyte adhesion positive regulation of pyloric antrum smooth muscle contraction positive regulation of colon smooth muscle contraction positive regulation of small intestine smooth muscle contraction positive regulation of vascular smooth muscle cell differentiation signal transduction stem cell population maintenance regulation of cell proliferation positive regulation of protein kinase B signaling mast cell proliferation germ cell development cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-74d7b4b4d9-zrl6d:80/100.66.77.38:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline