Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Description: This 20RNTC monoclonal antibody recognizes mouse and human IL-20R2 (interleukin-20 receptor 2, IL-20Rb) in either of its two heterodimeric complexes formed by pairing with IL-20R1 or IL-22R1. IL-20R2 belongs to the class II cytokine receptor family that shares a characteristic pattern of cysteine, proline and tryptophan residues in the extracellular domain. IL-20R2 paired with IL-20R1 forms a receptor that binds IL-20, IL-19, or IL-24. IL-20R2 paired with IL22R1 forms a receptor that binds IL-20 or IL-24. The IL-20R complex signals primarily via the JAK-STAT pathway. IL-20R2 is found predominantly on cells of epithelial origin such as gut and skin and can be upregulated during inflammation. Immunofluorescent staining of A375 cells reveals a punctate staining pattern at the cell membrane, as well as diffuse, cytoplasmic staining that is likely associated with protein synthesis and transport in the endoplasmic reticulum and golgi.
Applications Reported: This 20RNTC antibody has been reported for use in immunocytochemistry, immunoblotting (WB) and flow cytometric analysis.
Applications Tested: This 20RNTC antibody has been tested by immunofluorescent staining and flow cytometric analysis of fixed and permeabilized A375 cells. This can be used between 1-10 µg/mL.
Purity: Greater than 90%, as determined by SDS-PAGE.
Aggregation: Less than 10%, as determined by HPLC.
Filtration: 0.2 µm post-manufacturing filtered.
IL20RB and IL20RA (MIM 605620) form a heterodimeric receptor for interleukin-20 (IL20; MIM 605619) (Blumberg et al., 2001 [PubMed 11163236]).[supplied by OMIM, Feb 2009].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: fibronectin type III domain containing 6; IL-20 receptor subunit beta; IL-20R2/IL-20rb; interleukin 20 receptor beta subunit; interleukin-20 receptor II; unnamed protein product
Gene Aliases: DIRS1; FNDC6; Gm186; IL-20R-beta; IL-20R2; IL-20RB; Il20R2
UniProt ID: (Human) Q6UXL0
Entrez Gene ID: (Human) 53833, (Mouse) 213208
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support