Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This AS 4C5 monoclonal antibody recognizes human ASH2L, which has been described as a transcriptional regulator and may play a role in hematopoiesis. AS 4C5 detects full length (unspliced) and reduced ASH2L by Western Blot and stains ASH2L in the nucleoplasm and at the plasma membrane in Immunocytochemistry. ASH2L monoclonal antibody (AS 4C5) has been validated for the following applications: Western Blot and Immunocytochemistry; AS 4C5 does not cross react with mouse ASH2L.
Applications Reported: This AS 4C5 antibody has been reported for use in Western Blot and Immunocytochemistry.
Applications Tested: This AS 4C5 antibody has been tested by western blot using whole cell lysates of HeLa cells (human) and NIH 3T3 or N2a (mouse cell lines) and by immunocytochemistry on HeLa cells. This AS 4C5 antibody may be used at 2 µg/mL for western blot and at 1.25 µg/mL for immunocytochemistry.
The human ASH2L gene encodes a 628 amino acid protein known as ASH2L1, or isoform 1, which contains a nuclear localization signal and PHD finger motif, suggesting that the gene product functions as a transcription regulator. Its predicted molecular weight is 68.7kDa. Alternative splicing results in possible isoforms at 60 kDa & 56.5 kDa). Human ASH2L proteins are 60% homologous to Drosophila ash2, which positively regulates expression of certain genes in early development, and contain similar, but not identical, domains, including a zinc finger motif. Homology to mouse ASH2L is 90%. ASH2L is predominantly expressed in adult heart and testis and fetal lung and liver, with barely detectable expression in adult lung, liver, kidney, prostate, and peripheral leukocytes as well as leukemia cell lines with erythroid and megakaryocytic potential, such as K562, Hel and Dami. Differentiation inducers (e.g. phorbol ester and hemin) cause different expression patterns in these cell lines, suggesting that ASH2L plays a role in hematopoiesis and is associated with particular types of leukemia. In addition ASH2L has been suggested to interact with beta-tubulin and beta-catenin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ASH 2; ash2 (absent, small, or homeotic)-like; ASH2 LIKE; ASH2 like protein; ASH2-like protein; ASH2L; Bre 2; unnamed protein product
Gene Aliases: ASH2; ASH2L1; ASH2L2; Bre2
UniProt ID: (Human) Q9UBL3
Entrez Gene ID: (Human) 9070
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support