Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • IL-1 beta Antibodies

          Invitrogen

          IL-1 beta Monoclonal Antibody (CRM56), eBioscience™

          7 Published Figures
          17 References
          View all (144) IL-1 beta antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite IL-1 beta Monoclonal Antibody (CRM56), eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (7)
          IL-1 beta Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          IL-1 beta Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 8

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IL-1 beta Antibody (14-7018-81) in Flow

          Staining of human peripheral blood mononuclear cells stimulated with LPS in the presence of Brefeldin A for 4 hours with Anti-Human CD14 PE (Product # 12-0149-42) (left) or Anti-Human CD14 FITC (Product # 11-0149-42) (right) followed by intracellularly Anti-Human IL-1 beta PE. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IL-1 beta Antibody in Flow Cytometry (Flow)
          IL-1 beta Antibody in Flow Cytometry (Flow)
          IL-1 beta Antibody in Flow Cytometry (Flow)
          IL-1 beta Antibody in Flow Cytometry (Flow)
          IL-1 beta Antibody in Flow Cytometry (Flow)
          IL-1 beta Antibody in Flow Cytometry (Flow)
          IL-1 beta Antibody in Neutralization (Neu)
          IL-1 beta Antibody in Neutralization (Neu)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          14-7018-81

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          -
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          Assay-Dependent
          View 5 publications 5 publications

          ELISA (ELISA)

          1-4 µg/mL
          View 7 publications 7 publications

          Neutralization (Neu)

          Assay-Dependent
          View 3 publications 3 publications

          Functional Assay (Functional)

          Assay-Dependent
          -

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human

          Published species

          Human, Mouse

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          CRM56

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 647 
          • Brilliant UV 395
          • Brilliant Violet 421
          • Brilliant Violet 650
          • FITC
          • Functional Grade
          • PE 
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_468400

          Product Specific Information

          Description: The CRM56 antibody reacts with human and baboon interleukin-1 beta; (IL-1 beta). IL-1 beta is a 17 kDa factor secreted primarily by monocytes. IL-1 has effects on T-helper cells, inducing IL-2 secretion and expression of IL-2 receptors. IL-1 has effects on B cells, promoting cell proliferation and immunoglobulin synthesis.

          Applications Reported: The CRM56 antibody has been reported for use in intracellular staining for flow cytometric analysis, cytokine neutralization, and ELISA. Fluorochrome conjugated CRM56 is recommended for use in intracellular staining for flow cytometry. Functional Grade purified CRM56 (Product # 16-7018) is recommended for use in functional assays.

          Applications Tested: The CRM56 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of human interleukin-1 beta (IL-1 beta) in combination with the biotin CRM57 (Product # 13-7016-81) antibody for detection and recombinant human IL-1 beta (14-8018) as the standard. A suitable range of concentrations of this antibody for ELISA capture is 1 - 4 µg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 1000 pg/mL - 8 pg/mL should be included in each ELISA plate.

          The Functional Grade Purified CRM56 antibody has been tested for neutralization of IL-1 beta bioactivity.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Interleukin-1 beta (IL-1 beta) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 beta is synthesized in response to inflammatory stimuli as a 31 kDa inactive pro-form that accumulates in the cytosol. Cleavage of pro-IL-1 beta into the active 17 kDa protein requires the activation of inflammasomes, which are multi-protein complexes that respond to pathogens, stress conditions, and other danger signals. Inflammasome activation triggers the processing of the caspase-1 precursor into its active form, which in turn cleaves pro-IL-1 beta. IL-1 beta lacks a signal sequence peptide for classical ER/Golgi pathway and is secreted alongside caspase-1 via an alternate and incompletely understood mechanism. Although IL-1 beta is most often secreted in its active form, secretion of the uncleaved protein may be detectable under some biological conditions. IL-1 beta signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 alpha. IL-1 beta activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 beta play an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses to infection and injury. Elevated levels of IL-1 beta have been associated with many chronic inflammatory conditions IL-1 beta neutralizing antibodies potential therapeutic value.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: catabolin; Hematopoietin 1 (H1); IFN beta inducing factor; il 1b; IL 1β; IL-1 beta; IL1 B; IL1B; IL1β; ILN; Interleukin; Interleukin 1 beta precursor; interleukin 1 precursor polypeptide; interleukin 1beta; Interleukin-1b; Interleukin1 beta; LAF; Osteoclast activating factor (OAF); preinterleukin 1 beta; Pro interleukin 1 beta; pro-interleukin-1-beta; unnamed protein product

          View more View less

          Gene Aliases: IL-1; IL1-BETA; IL1beta; IL1F2

          View more View less

          UniProt ID: (Human) P01584

          View more View less

          Entrez Gene ID: (Human) 3553

          View more View less

          Function(s)
          cytokine activity interleukin-1 receptor binding integrin binding protein binding protein domain specific binding receptor agonist activity
          Process(es)
          positive regulation of cytokine production microglial cell activation involved in immune response positive regulation of T cell mediated immunity apoptotic process inflammatory response immune response signal transduction cell-cell signaling embryo implantation positive regulation of cell proliferation negative regulation of cell proliferation vascular endothelial growth factor production positive regulation of vascular endothelial growth factor production positive regulation of gene expression positive regulation of platelet-derived growth factor receptor signaling pathway positive regulation of epithelial to mesenchymal transition positive regulation of macrophage derived foam cell differentiation negative regulation of glucose transport smooth muscle adaptation cytokine-mediated signaling pathway hyaluronan biosynthetic process monocyte differentiation positive regulation of cell migration positive regulation of vascular endothelial growth factor receptor signaling pathway positive regulation of prostaglandin biosynthetic process positive regulation of fever generation positive regulation of prostaglandin secretion response to lipopolysaccharide negative regulation of interleukin-1 beta production positive regulation of granulocyte macrophage colony-stimulating factor production positive regulation of interferon-gamma production positive regulation of interleukin-2 production positive regulation of interleukin-6 production positive regulation of interleukin-8 production positive regulation of immature T cell proliferation in thymus positive regulation of heterotypic cell-cell adhesion positive regulation of T cell proliferation regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of I-kappaB kinase/NF-kappaB signaling negative regulation of MAPK cascade positive regulation of MAPK cascade positive regulation of nitric oxide biosynthetic process positive regulation of angiogenesis negative regulation of lipid metabolic process positive regulation of mitotic nuclear division positive regulation of transcription, DNA-templated positive regulation of complement activation positive regulation of JNK cascade negative regulation of insulin receptor signaling pathway positive regulation of protein export from nucleus regulation of defense response to virus by host negative regulation of inflammatory response positive regulation of inflammatory response regulation of neurogenesis negative regulation of neurogenesis regulation of insulin secretion negative regulation of synaptic transmission defense response to Gram-positive bacterium negative regulation of lipid catabolic process positive regulation of lipid catabolic process regulation of nitric-oxide synthase activity positive regulation of membrane protein ectodomain proteolysis positive regulation of protein kinase B signaling positive regulation of glial cell proliferation positive regulation of cell adhesion molecule production negative regulation of adiponectin secretion regulation of ERK1 and ERK2 cascade positive regulation of ERK1 and ERK2 cascade monocyte aggregation interleukin-1-mediated signaling pathway response to interleukin-1 cellular response to lipopolysaccharide cellular response to mechanical stimulus cellular response to xenobiotic stimulus positive regulation of monocyte chemotactic protein-1 production cellular response to interleukin-17 positive regulation of neuroinflammatory response positive regulation of p38MAPK cascade positive regulation of NIK/NF-kappaB signaling positive regulation of RNA biosynthetic process regulation of establishment of endothelial barrier regulation of secretion by cell negative regulation of gap junction assembly positive regulation of occluding junction disassembly positive regulation of T-helper 1 cell cytokine production negative regulation of interleukin-1-mediated signaling pathway negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-9vkmd:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0