Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 11
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Description: The TRA-1-81 antibody recognizes a protein expressed on undifferentiated human embryonic stem cells (ES), embryonic carcinoma cells (EC), and embryonic germ cells (EG). Like other stem cell specific markers, the epitope recognized by the TRA-1-81 antibody is lost upon cell differentiation. The TRA-1-81 epitope is resistant to neuraminidase digestion, unlike the epitope recognized by the related TRA-1-60 antibody. The TRA-1-81 antibody is known to specifically recognize a carbohydrate epitope on a keratan sulfated glycoprotein recently identified as podocalyxin, a member of the CD34-related family of sialomucins. Podocalyxin is a transmembrane glycoprotein originally identified on epithelial glomerular cells known as podocytes, and the protein has also been implicated in the development of aggressiveness in a variety of cancers, including breast and prostate cancer.
Applications Reported: This TRA-1-81 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, immunoblotting, and immunofluorescence.
Applications Tested: This TRA-1-81 antibody has been tested by flow cytometric analysis of the human embryonic carcinoma (EC) line 2102Ep. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. This antibody has also been tested by immunoblotting of 2102Ep cell extracts, yielding a band of approximately 200-250 kDa (reducing and non-reducing conditions). This antibody has been tested by immunofluorescent staining of formaldehyde fixed and permeabilized cells and can be used at less than or equal to 5µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Purity: Greater than 90%, as determined by SDS-PAGE.
Aggregation: Less than 10%, as determined by HPLC.
Filtration: 0.2 µm post-manufacturing filtered.
TRA-1-81 is a cell surface antigen expressed along with SSEA-3, SSEA-4 and TRA-1-60 in human embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). These surface markers are down-regulated during the differentiation process. In contrast, SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: tra1-81
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support