Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • New Products
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NGFR (CD271) Antibodies

          Invitrogen

          CD271 (NGF Receptor) Monoclonal Antibody (ME20.4), eBioscience™

          4 Published Figures
          7 References
          View all (102) NGFR (CD271) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD271 (NGF Receptor) Monoclonal Antibody (ME20.4), eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (4)
          CD271 (NGF Receptor) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          CD271 (NGF Receptor) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD271 (NGF Receptor) Antibody (14-9400-37) in IHC (P)

          Immunohistochemistry of formalin-fixed paraffin embedded human prostate tissue stained with 5 µg/mL of Mouse IgG1 K Isotype Control Purified (left) or 5 µg/mL of Anti-CD271 (NGF Receptor) Purified (right) followed by Anti-Mouse IgG Biotin, Streptavidin HRP, and DAB visualization.Nuclei are counterstained with hematoxylin. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD271 (NGF Receptor) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD271 (NGF Receptor) Antibody in Immunohistochemistry (IHC)
          CD271 (NGF Receptor) Antibody in Immunohistochemistry (IHC)
          CD271 (NGF Receptor) Antibody in Western Blot (WB)
          CD271 (NGF Receptor) Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          14-9400-37

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-Dependent
          View 2 publications 2 publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          5 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          Assay-Dependent
          -

          Flow Cytometry (Flow)

          0.25 µg/test
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Dog, Human, Mouse, Non-human primate, Sheep, Pig, Rabbit, Rat

          Published species

          Dog, Human, Mouse

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          ME20.4

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 488 
          • APC 
          • PE 
          • Super Bright 436
          • Super Bright 600
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          2 mg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2865377

          Product Specific Information

          Description: The monoclonal antibody ME20.4 recognizes CD271, a 75 kDa type I transmembrane protein that belongs to the low-affinity neurotrophin receptor and tumor necrosis factor receptor superfamily. CD271 is expressed by neurons, Schwann cells, melanocytes, mesenchymal stem/stromal cells, neural crest stem cells, astrocytes, oligodendrocytes, and a subset of lymphoid cells including follicular dendritic cells. CD271 binds to the ligands NGF, BDNF, NTF3, and NTF4. CD271 contains a death domain which induces apoptosis and regulates cell growth and proliferation through interactions with TRAF2, TRAF4, TRAF6, PTPN13, and RANBP9.

          The ME20.4 monoclonal antibody recognizes human, non-human primate, mouse, rat, sheep, rabbit, cat, dog, and pig CD271.

          Applications Reported: This ME20.4 antibody has been reported for use in flow cytometric analysis, western blotting, microscopy, immunohistochemical staining, and immunocytochemistry.

          Applications Tested: This ME20.4 antibody has been tested by flow cytometric analysis of SK-N-SH cells. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. This ME20.4 antibody has also been tested by immunohistochemistry of formalin-fixed paraffin embedded human tissue using high pH antigen retrieval and can be used at less than or equal to 5 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Neurotrophic factors control the survival, differentiation and maintenance of neurons in the peripheral and central nervous systems, and of other neural crest-derived cell types. Developing sympathetic neurons are absolutely dependent upon nerve growth factor (NGF) during the period of target competition in vivo. Antibodies reacting specifically with NGFR p75 are useful tools in the detection and characterization of NGFR p75, to enhance our understanding of a wide range of phenomena in the development, plasticity and repair of the nervous system.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CD271 antigen; gp80-LNGFR; low affinity nerve growth factor receptor; low affinity neurotrophin receptor p75NTR; low-affinity nerve growth factor receptor; low-affinity nerve growth factor receptor p75NGFR; low-affinity nerve growth factor receptor p75NGR; nerve growth factor receptor (TNFR superfamily, member 16); nerve growth factor receptor, fast; NGF receptor; NGFR; p-75; p75 ICD; p75 neurotrophin receptor; p75 ntr; TNFR superfamily, member 16; TNR16; unnamed protein product

          View more View less

          Gene Aliases: CD271; Gp80-LNGFR; LNGFR; NGR; p75; p75(NTR); p75NGFR; p75NTR; RNNGFRR; TNFRSF16

          View more View less

          UniProt ID: (Human) P08138, (Mouse) Q9Z0W1, (Rat) P07174

          View more View less

          Entrez Gene ID: (Pig) 100525607, (Human) 4804, (Sheep) 101119064, (Dog) 491071, (Rabbit) 100349095, (Mouse) 18053, (Rat) 24596

          View more View less

          Function(s)
          beta-amyloid binding transmembrane signaling receptor activity death receptor activity protein binding calmodulin binding coreceptor activity small GTPase binding ubiquitin protein ligase binding signaling receptor activity neurotrophin binding nerve growth factor binding neurotrophin TRKA receptor binding identical protein binding macromolecular complex binding preprotein binding transmembrane signal receptor
          Process(es)
          cellular glucose homeostasis intracellular protein transport signal transduction Rho protein signal transduction circadian rhythm negative regulation of cell migration circadian regulation of gene expression dorsal aorta development regulation of cell proliferation glucose homeostasis positive regulation of apoptotic process neuron apoptotic process extrinsic apoptotic signaling pathway presynaptic modulation of chemical synaptic transmission positive regulation of protein localization to nucleus positive regulation of pri-miRNA transcription from RNA polymerase II promoter negative regulation of blood vessel endothelial cell proliferation involved in sprouting angiogenesis cellular response to beta-amyloid positive regulation of endothelial cell apoptotic process axon guidance central nervous system development regulation of gene expression positive regulation of neuron projection development negative regulation of neuron projection development detection of temperature stimulus negative regulation of angiogenesis nerve development hair follicle morphogenesis positive regulation of myelination positive regulation of synaptic transmission, cholinergic cellular response to oxidative stress positive regulation of Rho protein signal transduction negative regulation of fibroblast growth factor receptor signaling pathway positive regulation of odontogenesis of dentin-containing tooth negative regulation of apoptotic process positive regulation of MAPK cascade negative regulation of neuron apoptotic process skin development positive regulation of neuron differentiation positive regulation of fibroblast proliferation negative regulation of multicellular organismal process negative regulation of hair follicle development positive regulation of protein kinase B signaling negative regulation of mitochondrial depolarization positive regulation of synaptic transmission, glutamatergic negative regulation of dendritic spine development positive regulation of neural precursor cell proliferation regulation of reactive oxygen species metabolic process positive regulation of excitatory postsynaptic potential positive regulation of apoptotic signaling pathway spermatogenesis response to lipopolysaccharide skeletal muscle cell differentiation response to ethanol
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-mwps6:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0