Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • FOXJ1 Antibodies

          Invitrogen

          FOXJ1 Monoclonal Antibody (2A5), eBioscience™

          Advanced Verification
          74 Published Figures
          123 References
          View all (14) FOXJ1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite FOXJ1 Monoclonal Antibody (2A5), eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          • Published Figures (74)
          • Advanced Verification (1)
          FOXJ1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FOXJ1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 77

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          FOXJ1 Antibody (14-9965-37) in IHC (P)

          Immunohistochemistry of paraffin-embedded mouse lung tissue with Anti-Human/Mouse FOXJ1 Purified followed by Anti-Mouse IgG Biotin (Product # 13-4013-85) and Avidin HRP (Product # 18-4100-51) (right).Data provided courtesy of Steven L.Brody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          FOXJ1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FOXJ1 Antibody in Western Blot (WB)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          FOXJ1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          FOXJ1 Antibody in Immunohistochemistry (Frozen), Immunocytochemistry (IHC (F), ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry (Frozen), Immunocytochemistry (IHC (F), ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry, Immunohistochemistry (PFA fixed), Immunocytochemistry (IHC, IHC (PFA), ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FOXJ1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FOXJ1 Antibody in Immunohistochemistry, Immunohistochemistry (Frozen) (IHC, IHC (F))
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          FOXJ1 Antibody in Western Blot (WB)
          FOXJ1 Antibody in Western Blot, ChIP Assay (WB, ChIP)
          FOXJ1 Antibody in Western Blot (WB)
          FOXJ1 Antibody in Western Blot (WB)
          FOXJ1 Antibody in Western Blot (WB)
          FOXJ1 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          14-9965-37

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-10 µg/mL
          View 6 publications 6 publications

          Immunohistochemistry (IHC)

          -
          View 64 publications 64 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1-10 µg/mL
          View 6 publications 6 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 3 publications 3 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 5 publications 5 publications

          Immunocytochemistry (ICC/IF)

          Assay-Dependent
          View 35 publications 35 publications

          Flow Cytometry (Flow)

          -
          View 2 publications 2 publications

          ChIP assay (ChIP)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Fruit fly, Human, Mouse, Pig, Rat, Zebrafish

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          2A5

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          2 mg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2865539

          Product Specific Information

          Description: This 2A5 monoclonal antibody reacts with Foxj1, a 50-kDa member of the Forkhead-box (FOX) family of winged-helix transcription factors. Expression of Foxj1 is high in ciliated epithelial cells and low in naive T and B cells, where cellular activation results in downregulation of the factor. Foxj1 modulates germinal center formation and Th1 activation by repressing NFkappaB through IkappaBbeta induction. As such, this transcription factor has been suggested to antagonize autoimmune reactions such as systemic lupus erythematosus and rheumatoid arthritis. In addition, Foxj1 has been linked to the formation of motile cilia in epithelial cells.

          Applications Reported: This 2A5 antibody has been reported for use in western blotting, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and immunocytochemistry.

          Applications Tested: This 2A5 antibody has been tested by western blot of mouse tracheal epithelial cells and testes, as well as by immunohistochemistry of mouse lung tissue. This antibody can be used between 1-10 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Forkhead-box J1 (FOXJ1) is a 421-amino acid transcription factor that suppresses T cell activity and spontaneous autoimmunity, through the repression of NF B activity. FOXJ1 also inhibits the humoral immune response in B cells. FOXJ1 deficiency in B cells results in spontaneous and accentuated germinal center formation and is implicated in the development of pathogenic autoantibodies and accentuated responses to immunizations. Abnormal expression of FOXJ1 may be associated with autoimmune diseases and/or other inflammatory diseases. FOXJ1 is also required for cilia formation and left-right axis determination because it increases calpastatin expression, a protein necessary for the ability of basal bodies to anchor to the apical cytoskeleton. FOXJ1 expression may function as an early marker of epithelial cell differentiation, recovery, and function.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: fork head homologue 4; forkhead transcription factor HFH-4; forkhead-like 13; forkhead-related protein FKHL13; hepatocyte nuclear factor 3 forkhead homolog 4; HNF-3/fork-head homolog-4; HNF-3/forkhead homolog 4; MGC35202; unnamed protein product

          View more View less

          Gene Aliases: CILD43; FKHL-13; FKHL13; HFH-4; HFH4

          View more View less

          UniProt ID: (Human) Q92949, (Mouse) Q61660

          View more View less

          Entrez Gene ID: (Human) 2302, (Mouse) 15223

          View more View less

          Function(s)
          transcription regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding protein binding sequence-specific DNA binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter central tolerance induction negative regulation of germinal center formation positive regulation of central B cell tolerance induction negative regulation of humoral immune response mediated by circulating immunoglobulin regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter humoral immune response determination of left/right symmetry brain development heart development protein localization gene expression actin cytoskeleton organization regulation of epithelial cell differentiation ciliary basal body organization negative regulation of interleukin-6 production negative regulation of T cell differentiation in thymus axoneme assembly establishment of apical/basal cell polarity metanephric part of ureteric bud development negative regulation of T cell proliferation motile cilium assembly positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter cell maturation negative regulation of B cell activation leukocyte migration cilium morphogenesis lung epithelium development epithelium development left/right pattern formation glomerular parietal epithelial cell development negative regulation of NIK/NF-kappaB signaling positive regulation of lung ciliated cell differentiation spermatogenesis pattern specification process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-d6wk2:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0