What is the difference between DPBS, no calcium, no magnesium, 10 x 500 mL, Cat. Nos. 14190250 and 1490169?
DPBS, no calcium, no magnesium, 10x 500 mL (Cat. Nos. 14190250 and 14190169) have the same formulation. The only difference is in the manufacturing site.
The DPBS, no calcium, no magnesium (Cat. no. 14190169) is manufactured in our Inchinnan, Scotland facility and is intended for sale in Europe, Africa, and the Middle East—higher freight charges will apply to orders shipped elsewhere. DPBS, no calcium, no magnesium (Cat. No. 14190250) is manufactured in Grand Island, US, and is recommended for our customers in North America, Latin America, and the Asia/Pacific region.
The complete formulation can be found here:
14190 - DPBS, no calcium, no magnesium
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
The DPBS, no calcium, no magnesium (Cat. no. 14190169) is manufactured in our Inchinnan, Scotland facility and is intended for sale in Europe, Africa, and the Middle East—higher freight charges will apply to orders shipped elsewhere. DPBS, no calcium, no magnesium (Cat. No. 14190250) is manufactured in Grand Island, US, and is recommended for our customers in North America, Latin America, and the Asia/Pacific region.
The complete formulation can be found here:
14190 - DPBS, no calcium, no magnesium
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.