Search
| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Inorganic Salts | |||
| Potassium Chloride (KCl) | 75.0 | 200.0 | 2.6666667 |
| Potassium Phosphate monobasic (KH2PO4) | 136.0 | 200.0 | 1.4705882 |
| Sodium Chloride (NaCl) | 58.0 | 8000.0 | 137.93103 |
| Sodium Phosphate dibasic (Na2HPO4-7H2O) | 268.0 | 2160.0 | 8.059702 |
1. Dulbecco, R. and Vogt, M., (1954) Plaque formation and isolation of pure lines with Poliomyelitis viruses. J. Exp. Med., 98:167.
14200 - DPBS, 10x, no calcium, no magnesium
Cat No:
14200059
,
14200067
,
14200075
,
14200083
,
14200091
,
14200166
21600 - DPBS, powder, no calcium, no magnesium
Cat No:
21600010
,
21600044
,
21600069