3-Diethylaminophenol, 99%
3-Diethylaminophenol, 99%
3-Diethylaminophenol, 99%
Thermo Scientific Chemicals

3-Diethylaminophenol, 99%

CAS: 91-68-9 | C10H15NO | 165.24 g/mol
Have Questions?
Catalog NumberQuantity
146631000100 g
Catalog number 146631000
Price (EUR)
37,80
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
37,80
Each
Chemical Identifiers
CAS91-68-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Flakes or granules
Appearance (Color)Pink to red to grey-brown
Infrared spectrumConforms
GC>=98.5 %
Melting point70°C to 73°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only