2,4-Dichlorophenol, 99%
2,4-Dichlorophenol, 99%
2,4-Dichlorophenol, 99%
Thermo Scientific Chemicals

2,4-Dichlorophenol, 99%

CAS: 120-83-2 | C6H4Cl2O | 163.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 147721000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS120-83-2
IUPAC Name2,4-dichlorophenol
Molecular FormulaC6H4Cl2O
InChI KeyHFZWRUODUSTPEG-UHFFFAOYSA-N
SMILESOC1=CC=C(Cl)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline solid
Infrared spectrumConforms
Melting point41°C to 44°C
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only