QuantaRed™ Enhanced Chemifluorescent HRP Substrate Kit
Thermo Scientific™

QuantaRed™ Enhanced Chemifluorescent HRP Substrate Kit

The Thermo Scientific QuantaRed Enhanced Chemifluorescent HRP Substrate (ADHP) is the most sensitive fluorescent ELISA substrate, providing sensitivity comparable toRead more
Have Questions?
Catalog NumberQuantity
15159110 ml
Catalog number 15159
Price (USD)
518.00
Each
Quantity:
110 ml
Request bulk or custom format
Price (USD)
518.00
Each
The Thermo Scientific QuantaRed Enhanced Chemifluorescent HRP Substrate (ADHP) is the most sensitive fluorescent ELISA substrate, providing sensitivity comparable to enhanced chemiluminescence. This kit contains QuantaRed ADHP concentrate, QuantaRed Enhancer Solution, QuantaRed Stable Peroxide Solution, and QuantRed Stop Solution, and is sufficient for 960 wells (10 x 96-well plates at 100 μL working solution per well.

Features of the QuantaRed Enhanced Chemifluorescent HRP Substrate Kit:

• Sensitive—low picogram to femtogram detection
• Red-Shifted - minimal autofluorescence interference from biological samples
• Convenient—compatible with existing filter sets, no need to purchase special filter sets
• Complete—mix and use reagents, no buffers to prepare or reagents to dissolve
• Flexible—use Stop Solution to effectively stop the reaction resulting in stable signal for at least 4 hours or more.
• Versatile—produces a strong colorimetric signal and can be read fluorescently or colorimetrically at 570nm

QuantaRed HRP Substrate enables high-yield and sensitive fluorescent and colorimetric detection of peroxidase activity in the ELISA format. QuantaRed Substrate uses a proprietary technology to increase fluorescent yield and sensitivity of the well-documented ADHP (10-Acetyl-3,7-dihydroxyphenoxazine) chemifluorescence reaction. ADHP is a non-fluorescent compound that reacts with horseradish peroxidase (HRP) to produce resorufin, a soluble, highly fluorescent reaction product with excitation/emission maxima of ∼570/585nm. The long wavelength emission of the resorufin minimizes interference from the low wavelength (blue and green) autofluorescence, making it an ideal substrate for quantitating target molecules in biological samples. In addition, the reaction product of QuantaRed Substrate is colorimetric as well as being fluorescent and can be read spectrophotmetrically at ∼570nm.

Specially formulated, QuantaRed Substrate incorporates chemical enhancers for maximum sensitivity and superior low end linearity. With high quantum yield and large extinction coefficient, QuantaRed Enhanced HRP Substrate has sensitivity comparable to enhanced chemiluminescence and is more sensitive than the Amplex Red substrates for the detection of HRP in ELISA applications.
WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Product LineQuantaRed
Substrate PropertiesChemical Substrate
Substrate TypeHRP (Horseradish Peroxidase)
Product TypeELISA Substrate
Quantity110 ml
Unit SizeEach
Contents & Storage
Sufficient For: 960 wells (10 x 96-well plates) at 100 μL working solution per well
• QuantaRed ADHP Concentrate, 1 mL
• QuantaRed Enhancer Solution, 50 mL
• QuantaRed Stable Peroxide Solution, 50 mL
• QuantaRed Stop Solution, 10 mL

Store at 4°C.

Frequently asked questions (FAQs)

Can QuantaRed Enhanced Chemifluorescent HRP Substrate be used for Western blotting?.

No. This is a soluble HRP substrate for ELISA and other plate-based applications.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What antibody concentrations are recommended for use with QuantaRed Enhanced Chemifluorescent HRP Substrate?

The recommended concentrations of antibodies and HRP conjugate are listed below:

Capture Antibody: 1-10 µg/mL
Detection Antibody: 0.05-1 µg/mL
HRP Conjugate: 0.1-0.2 µg/mL

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Can a stop solution from another source be used with the QuantaRed Enhanced Chemifluorescent HRP Substrate?

No. The QuantaRed Stop Solution included in the kit must be used to effectively stop the reaction.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Can the QuantaRed Enhanced Chemifluorescent HRP Substrate signal be measured in a kinetic mode without stopping the reaction?

To measure in a kinetic mode, simply omit the Stop Solution. At high HRP concentrations and prolonged reaction times, the fluorescent product may self-quench, which is observed as continuous color development without a change in fluorescent intensity. Optimization of the detection antibody or HRP conjugate is required to determine the linear range of a particular assay system.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What types of plates are compatible with the QuantaRed Enhanced Chemifluorescent HRP Substrate?

Black opaque plates typically produce the best results. White opaque plates also can be used, but might produce higher backgrounds. A variety of other plate types, such as gray or opaque plates with transparent bottoms are also useful with this substrate. Use clear plates if the absorbance of the colored product will be measure instead of the fluorescence.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.