QuantaRed™ Enhanced Chemifluorescent HRP Substrate Kit - FAQs
QuantaRed™ Enhanced Chemifluorescent HRP Substrate Kit - FAQs
View additional product information for QuantaRed™ Enhanced Chemifluorescent HRP Substrate Kit - FAQs (15159)
10 product FAQs found
Can QuantaRed Enhanced Chemifluorescent HRP Substrate be used for Western blotting?.
No. This is a soluble HRP substrate for ELISA and other plate-based applications.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What antibody concentrations are recommended for use with QuantaRed Enhanced Chemifluorescent HRP Substrate?
The recommended concentrations of antibodies and HRP conjugate are listed below:
Capture Antibody: 1-10 µg/mL
Detection Antibody: 0.05-1 µg/mL
HRP Conjugate: 0.1-0.2 µg/mL
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Can a stop solution from another source be used with the QuantaRed Enhanced Chemifluorescent HRP Substrate?
No. The QuantaRed Stop Solution included in the kit must be used to effectively stop the reaction.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Can the QuantaRed Enhanced Chemifluorescent HRP Substrate signal be measured in a kinetic mode without stopping the reaction?
To measure in a kinetic mode, simply omit the Stop Solution. At high HRP concentrations and prolonged reaction times, the fluorescent product may self-quench, which is observed as continuous color development without a change in fluorescent intensity. Optimization of the detection antibody or HRP conjugate is required to determine the linear range of a particular assay system.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What types of plates are compatible with the QuantaRed Enhanced Chemifluorescent HRP Substrate?
Black opaque plates typically produce the best results. White opaque plates also can be used, but might produce higher backgrounds. A variety of other plate types, such as gray or opaque plates with transparent bottoms are also useful with this substrate. Use clear plates if the absorbance of the colored product will be measure instead of the fluorescence.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Can the QuantaRed Enhanced Chemifluorescent HRP Substrate be used colorimetrically?
Yes. The color produced by the reaction is dark magenta and can be measured at 570 +/-10 nm on a colorimetric plate reader.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What type of instrumentation and filter sets are needed for quantifying the QuantaRed Enhanced Chemifluorescent HRP Substrate signal?
The QuantaRed Signal can be measured using standard fluorometers equipped with Cy3 Dye filter sets. Quantitation does not require filters that precisely match the excitation/emission maxima; however, a non-overlapping filter set with a bandpass that includes the excitation/emission spectra is required.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Can other wavelengths for excitation and emission be used to quantify HRP using the QuantaRed Enhanced Chemifluorescent HRP Substrate?
Yes. Wavelengths at 530-575 nm for excitation and 585-630 nm for emission can be used for signal detection.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What is included in the QuantaRed Enhanced Chemifluorescent HRP Substrate Kit?
The kit includes QuantaRed ADHP Concentrate, Enhancer Solution, Stable Peroxide Solution and Stop Solution.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What is the QuantaRed Enhanced Chemifluorescent HRP Substrate?
QuantaRed Enhanced Chemifluorescent HRP Substrate is a proprietary formulation that uses chemical enhancers to increase fluorescence yield and sensitivity of 10-acetyl-3,7-dihydroxyphenoxazine (ADHP) chemifluorescence in plate-based (e.g., ELISA) assays. ADHP is a non-fluorescent compound that reacts with horseradish peroxidase (HRP) in the presence of peroxide to produce resorufin, a soluble, highly fluorescent compound with excitation/emission maxima at 570/585 nm. Unlike chemiluminescent substrates, the QuantaRed Enhanced Chemifluorescent HRP Substrate reaction can be stopped with the fluorescent signal remaining stable for several hours. The long wavelength emission spectrum of resorufin helps minimize interference from lower wavelength autofluorescence (blue and green) that exists in many biological samples.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.